Skip to Content
Merck
All Photos(1)

Key Documents

PHR1400

Supelco

Mometasone Furoate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Mometasone furoate, 9α,21-dichloro-11β-hydroxy-16α-methyl 3,20-dioxopregna-1,4-dien-17-yl furan-2-carboxylate, (11β,16α)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30Cl2O6
CAS Number:
Molecular Weight:
521.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 768
traceable to Ph. Eur. M2900000
traceable to USP 1445470

API family

mometasone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5ccco5)C(=O)CCl

InChI

1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1

InChI key

WOFMFGQZHJDGCX-ZULDAHANSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Mometasone Furoate is a medication with anti-inflammatory and antipruritic properties, commonly used to treat mixed infections of the skin. It is also used as a corticosteroid for the treatment of asthma and its mode of action involves reducing the inflammation of the airways and preventing the loss of lung functions.

Application

Mometasone Furoate may be used as a secondary pharma standard for the quantification of the analyte in pharmaceutical formulations using various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Mometasone furoate is an anti-inflammatory glucocorticoid.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3330 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validated high-performance thin-layer chromatographic (HPTLC) method for simultaneous determination of nadifloxacin, mometasone furoate, and miconazole nitrate cream using fractional factorial design
Patel GK, et al.
Journal of food and drug analysis, 24(3), 610-619 (2016)
Simultaneous estimation of Nadifloxacin and Mometasone Furoate in topical cream by HPTLC method
Kulkarni AA, et al.
Der Pharma Chemica, 2(3), 25-30 (2010)
Validated chromatographic methods for the simultaneous determination of Mometasone furoate and Formoterol fumarate dihydrate in a combined dosage form
Merey AH, et al.
Bulletin of Faculty of Pharmacy, Cairo University , 54(1), 99-106 (2016)
Anthony D'Urzo
Expert opinion on pharmacotherapy, 8(16), 2871-2884 (2007-10-25)
Mometasone furoate dry-powder inhaler (MF-DPI) is an inhaled corticosteroid (ICS) used for the treatment of persistent asthma in patients aged >or= 12 years. MF-DPI has low systemic bioavailability and high glucocorticoid receptor affinity compared with most other ICSs and modifies
Claudine M Baldwin et al.
Drugs, 68(12), 1723-1739 (2008-08-07)
Mometasone furoate (Nasonex) is a high-potency intranasal corticosteroid available for the treatment and/or prophylaxis of the nasal symptoms of seasonal allergic rhinitis (SAR) and perennial allergic rhinitis (PAR). In the EU, it is approved for use in patients aged >

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service