Skip to Content
Merck
All Photos(1)

Key Documents

PHR1369

Supelco

Thiamine pyrophosphate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Thiamine pyrophosphate, Aneurinepyrophosphoric acid, Cocarboxylase, Thiamine pyrophosphate chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19ClN4O7P2S
CAS Number:
Molecular Weight:
460.77
Beilstein:
3875902
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1656002

API family

thiamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

CC1=C(CCOP(OP(O)(O)=O)(O)=O)SC=[N+]1CC2=C(N)N=C(C)N=C2.[Cl-]

InChI

1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H

InChI key

YXVCLPJQTZXJLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Thiamine pyrophosphate is a coenzyme of transketolase that catalyzes the cleavage of ribulose-5-phosphate; thereby forming D-glyceraldehyde-3-phosphate. This reaction requires the addition of an acceptor aldehyde such as ribose-5-phosphate, glyceraldehye or glycolaldehyde.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1473 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Cressina et al.
Biochemical Society transactions, 39(2), 652-657 (2011-03-25)
Riboswitches are regions of mRNA to which a metabolite binds in the absence of proteins, resoulting in alteration of transcription, translation or splicing. The most widespread forms of riboswitches are those responsive to TPP (thiamine pyrophosphate) the active form of
Sachin Rustgi et al.
PloS one, 9(6), e100998-e100998 (2014-06-27)
Extensive application of imidazolinone (IMI) herbicides had a significant impact on barley productivity contributing to a continuous decline in its acreage over the last two decades. A possible solution to this problem is to transfer IMI-resistance from a recently characterized
[Mechanism of action of thiamine pyrophosphate enzymes].
G A Kochetov
Uspekhi sovremennoi biologii, 61(1), 17-31 (1966-01-01)
Nia L Marrott et al.
The Biochemical journal, 463(3), 405-412 (2014-08-05)
The four-component polypeptides of the 2-oxoacid dehydrogenase complex from the thermophilic archaeon Thermoplasma acidophilum assemble to give an active multienzyme complex possessing activity with the branched-chain 2-oxoacids derived from leucine, isoleucine and valine, and with pyruvate. The dihydrolipoyl acyl-transferase (E2)
Thiamine pyrophosphate-catalyzed enzymatic decarboxylation of -oxo acids.
J Ullrich et al.
Vitamins and hormones, 28, 365-398 (1970-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service