Skip to Content
Merck
All Photos(1)

Key Documents

PHR1200

Supelco

p-Chloroaniline

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Chloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
471359
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1111908

vapor density

4.4 (vs air)

vapor pressure

0.15 mmHg ( 25 °C)

API family

proguanil, chlorhexidine 

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

232 °C (lit.)

mp

67-70 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(Cl)cc1

InChI

1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

QSNSCYSYFYORTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

p-Chloroaniline may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[1][2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3328 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and evaluation of lidocaine hydrochloride in cyclodextrin inclusion complexes for development of stable gel in association with chlorhexidine gluconate for urogenital use
da Silva LFJS, et al.
International journal of nanomedicine, 6(3), 1143-1143 (2011)
HPLC determination of chlorhexidine gluconate and p-chloroaniline in topical ointment
Havlikova L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 1169-1173 (2007)
Fabio Gosetti et al.
Environmental pollution (Barking, Essex : 1987), 158(2), 592-598 (2009-09-22)
This paper studies the degradation reactions that 4-chloroaniline can naturally undergo in waters for the action of sun light. 10.00 mg L(-1) 4-chloroaniline aqueous solution, without any addition of organic solvent, are undergone to photoirradiation under conditions that simulate sun
D Mortenson et al.
International endodontic journal, 45(9), 878-882 (2012-04-11)
To determine if the formation of para-chloroaniline (PCA) can be avoided by using an alternative irrigant following sodium hypochlorite but before chlorhexidine. Fifty-five single-rooted teeth were decoronated, instrumented to size 40, .06 taper whilst being irrigated with 14% ethylene-diamine-tetra-acetic acid
Kun Yang et al.
Environmental science & technology, 44(8), 3021-3027 (2010-03-06)
Competitive adsorption between nonpolar organic compounds and polar ionic organic compounds (IOCs) on carbon nanotubes (CNTs) is essential for application of CNTs as superior sorbents and for environmental risk assessment of both CNTs and organic contaminants. It was observed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service