Skip to Content
Merck
All Photos(1)

Documents

35823

Supelco

4-Chloroaniline

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
471359
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.4 (vs air)

vapor pressure

0.15 mmHg ( 25 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

232 °C (lit.)

mp

67-70 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(Cl)cc1

InChI

1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

QSNSCYSYFYORTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloroaniline is a chlorinated aromatic amines, which can generally find applications in industry in the production of cosmetics, dyes, herbicides and pharmaceutical products. It is also a major metabolite of the phenyl-urea herbicides.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron (III)-photoinduced degradation of 4-chloroaniline in aqueous solution
Mailhot G, et al.
Applied Catalysis. B, Environmental, 50, 25-35 (2004)
Biodegradation of 4-chloroaniline by bacteria enriched from soil
Vangnai.SA and Petchkroh W
FEMS Microbiology Letters, 268, 209-216 (2007)
Kun Yang et al.
Environmental science & technology, 42(21), 7931-7936 (2008-11-27)
Aqueous adsorption of a series of phenols and anilines by a multiwalled carbon nanotube material (MWCNT15), which depends strongly on the solution pH and the number and types of solute groups, was investigated in this study. The pH-dependent adsorption coefficients
Kun Yang et al.
Environmental science & technology, 44(8), 3021-3027 (2010-03-06)
Competitive adsorption between nonpolar organic compounds and polar ionic organic compounds (IOCs) on carbon nanotubes (CNTs) is essential for application of CNTs as superior sorbents and for environmental risk assessment of both CNTs and organic contaminants. It was observed in
Brian J Rasimick et al.
Journal of endodontics, 34(12), 1521-1523 (2008-11-26)
The combination of chlorhexidine and EDTA produces a white precipitate. The aim of this study was to determine if the precipitate involves the chemical degradation of chlorhexidine. The precipitate was produced and redissolved in a known amount of dilute trifluoroacetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service