Skip to Content
Merck
All Photos(1)

Documents

PHR1058

Supelco

Clotrimazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 379
traceable to Ph. Eur. C2430000
traceable to USP 1141002

API family

clotrimazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4

InChI

1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H

InChI key

VNFPBHJOKIVQEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Clotrimazole is an imidazole antifungal agent with relatively low toxicity. It is widely used in dermatology and gynaecology applications. Clotrimazole ointments are extensively utilized for the treatment of a wide range of systemic mycoses including candidiasis.

Application

Clotrimazole may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7129 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of HPLC method for determination of clotrimazole and its two degradation products in spray formulation
Hajkova R, et al.
Talanta, 73(3), 483-489 (2007)
Determination of methylparaben, propylparaben, clotrimazole and its degradation products in topical cream by RP-HPLC
Solich P, et al.
Chromatographia, 56(1), S181-S184 (2002)
Maria F Sassano et al.
Journal of medicinal chemistry, 56(6), 2406-2414 (2013-02-27)
Colloidal aggregation is the dominant mechanism for artifactual inhibition of soluble proteins, and controls against it are now widely deployed. Conversely, investigating this mechanism for membrane-bound receptors has proven difficult. Here we investigate the activity of four well-characterized aggregators against
Chromatographic determination of clotrimazole, ketoconazole and fluconazole in pharmaceutical formulations
Abdel-Moety EM, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 57(11), 931-938 (2002)
Jenna Corcoran et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 57-65 (2014-04-15)
Clotrimazole (CTZ) is a persistent imidazole antifungal agent which is frequently detected in the aquatic environment and predicted to bio-concentrate in fish. Common carp (Cyprinus carpio) were exposed to mean measured concentrations of either 1.02 or 14.63μgl(-1) CTZ for 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service