Skip to Content
Merck
All Photos(1)

Key Documents

P0550000

Perphenazine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26ClN3OS
CAS Number:
Molecular Weight:
403.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

perphenazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC2=C1SC3=CC=CC=C3N2CCCN4CCN(CCO)CC4

InChI

1S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2

InChI key

RGCVKNLCSQQDEP-UHFFFAOYSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Perphenazine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

D2 dopamine receptor antagonist; α-adrenergic receptor antagonist and σ-receptor agonist; phenothiazine antipsychotic. Inhibits glutamate dehydrogenase in vitro.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A David et al.
The Cochrane database of systematic reviews, (3)(3), CD001717-CD001717 (2005-07-22)
Antipsychotic drugs are usually given orally but compliance with medication given by this route may be difficult to quantify. The development of depot injections in the 1960s gave rise to extensive use of depots as a means of long-term maintenance
Disposition of the neuroleptics perphenazine, zuclopenthixol, and haloperidol cosegregates with polymorphic debrisoquine hydroxylation.
L Bertilsson et al.
Psychopharmacology series, 10, 230-237 (1993-01-01)
B Hartung et al.
The Cochrane database of systematic reviews, (1)(1), CD003443-CD003443 (2005-01-28)
Perphenazine is an old phenothiazine antipsychotic with a potency similar to haloperidol. It has been used for many years and is popular in the northern European countries and Japan. To examine the clinical effects and safety of perphenazine for those
S Quraishi et al.
The Cochrane database of systematic reviews, (2)(2), CD001717-CD001717 (2000-05-05)
Anti-psychotic drugs are usually given orally but compliance with medication given by this route may be difficult to quantify. The development of depot injections in the 1960s gave rise to extensive use of depots as a means of long-term maintenance
Perphenazine serum levels in patients on standard doses.
F Haffner
Psychopharmacology series, 7, 280-283 (1989-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service