Skip to Content
Merck
All Photos(1)

Documents

P0510000

Pentoxifylline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,7-Dimethyl-1-(5-oxohexyl)xanthine, Trental

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N4O3
CAS Number:
Molecular Weight:
278.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pentoxifylline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1C=NC2C1C(=O)N(CCCCC(C)=O)C(=O)N2C

InChI

1S/C13H20N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8,10-11H,4-7H2,1-3H3

InChI key

MQGNNJQTNFHNHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pentoxifylline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Phosphodiesterase inhibitor; inhibits synthesis of tumor necrosis factor α (TNF-α).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umair Sohail et al.
Clinics in liver disease, 16(4), 717-736 (2012-10-30)
Alcoholic hepatitis is a form of severe, cholestatic liver disease that results from consumption of large amount of alcohol during a sustained period of time in a subset of alcoholics. Symptoms could be mild and nonspecific to more severe. The diagnosis
Alejandro Bravo-Cuellar et al.
Journal of biomedical science, 20, 13-13 (2013-03-01)
In Oncology, the resistance of the cancerous cells to chemotherapy continues to be the principal limitation. The nuclear factor-kappa B (NF-κB) transcription factor plays an important role in tumor escape and resistance to chemotherapy and this factor regulates several pathways
Saad Hamama et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 105(3), 305-312 (2012-10-02)
Radiation-induced fibrosis is a serious late complication of radiotherapy. Pentoxifylline-vitamin E has proven effective and safe in clinical trials in the treatment of fibrosis, while the molecular mechanism of its activity is yet unexplored. Ten patients suffering from radiation-induced enteropathy
John M Akudugu et al.
International journal of radiation biology, 89(6), 462-470 (2013-02-01)
The mode by which the xanthine derivative, pentoxifylline, induces a radiosensitizing effect in cell cultures is a key and controversial radiobiological issue and requires further elucidation. Six human glioblastoma cell lines were tested for the effect of pentoxifylline treatment at
Simone Appenzeller et al.
Rheumatology international, 31(11), 1511-1513 (2009-12-17)
Behcet's disease is an inflammatory disorder of unknown cause, characterized by recurrent oral aphthous ulcers, genital ulcers, uveitis, and skin lesions. All these common manifestations are self-limiting except for the ocular attacks. The objective of the present study is to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service