Merck
  • Radiation-induced enteropathy: molecular basis of pentoxifylline-vitamin E anti-fibrotic effect involved TGF-β1 cascade inhibition.
Radiation-induced enteropathy: molecular basis of pentoxifylline-vitamin E anti-fibrotic effect involved TGF-β1 cascade inhibition.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology (2012-10-02)
Saad Hamama, Marie Gilbert-Sirieix, Marie-Catherine Vozenin, Sylvie Delanian
ABSTRACT
Radiation-induced fibrosis is a serious late complication of radiotherapy. Pentoxifylline-vitamin E has proven effective and safe in clinical trials in the treatment of fibrosis, while the molecular mechanism of its activity is yet unexplored. Ten patients suffering from radiation-induced enteropathy were treated with pentoxifylline-vitamin E combination with SOMA score as the primary endpoint. In parallel, primary smooth muscle cells isolated from intestinal samples isolated from humans with radiation enteropathy were incubated with pentoxifylline, trolox (vit. E hydrophilic analogous) or their combination. Activation of the TGF-β1/Smad and Rho/ROCK pathways was subsequently investigated using Q-RT-PCR, gene reporter, Western-blot, ELISA and immunohistochemistry. Pentoxifylline-vitamin E combination induces regression of symptoms (SOMA) by -41% and -80% at 6 and 18months. In vitro, pentoxifylline and trolox synergize to inhibit TGF-β1 protein and mRNA expression. This inhibitory action is mediated at the transcriptional level and leads to subsequent inhibition of TGF-β1/Smad targets (Col Iα1, FN1, PAI-1, CTGF), while it has no effect on the Rho/ROCK pathway. The anti-fibrotic effect of combined pentoxifylline-vitamin E is at least in part mediated by inhibition of the TGF-β1 cascade. It strengthens previous clinical data showing pentoxifylline-vitamin E synergy and supports its use as a first-line treatment of radiation-induced fibrosis.
Product Description
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pentoxifylline, solid