Skip to Content
Merck
All Photos(2)

Key Documents

NIST1474B

Polyethylene Resin

NIST®SRM®

Synonym(s):

Polyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

packaging

pkg of 60 g

manufacturer/tradename

NIST®

application(s)

chemicals and industrial polymers

format

neat

storage temp.

15-25°C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Each unit of SRM 1474b contains ~ 60 g of white polyethylene pellets packaged in an amber glass bottle. For more information, please refer to the SDS and COA.


SRM 1474B_cert

SRM 1474B_SDS

Application

The Standard Reference Material (SRM) Polyethylene Resin is primarily intended for calibrating and evaluating the performance of instruments in polymer technology and science, specifically for determining the melt flow rate according to ASTM D1238-13, Test Method for Flow Rates of Thermoplastics by Extrusion Plastometer, Standard Test Condition 190/2.16.

Features and Benefits

  • Certified values for melt flow rate were estimated using a Gaussian random effects model and the DerSimonian Laird procedure.
  • Expiration details and instructions for storage are provided on the NIST certificate.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service