Skip to Content
Merck
All Photos(1)

Documents

N0550000

Neostigmine methyl sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N2O6S
CAS Number:
Molecular Weight:
334.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

neostigmine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COS([O-])(=O)=O.CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

InChI

1S/C12H19N2O2.CH4O4S/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;1-5-6(2,3)4/h6-9H,1-5H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

OSZNNLWOYWAHSS-UHFFFAOYSA-M

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Neostigmine methyl sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Reversible inhibitor of acetylcholinesterase that is similar to eserine, but does not cross the blood-brain barrier.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsufumi Sato et al.
Masui. The Japanese journal of anesthesiology, 62(1), 19-26 (2013-02-26)
Anticholinesterase, such as neostigmine, was used to be a standard drug at the end of surgery for reversal of nondepolarizing neuromuscular block. Neostigmine decreases the metabolism of acetylcholine (ACh) at the neuromuscular junction and allows its concentration to increase and
Jakub Fichna et al.
Pharmacological reports : PR, 64(5), 1146-1154 (2012-12-15)
Animal models of visceral pain have gained much attention as an important tool to elucidate the possible mechanisms underlying functional gastrointestinal (GI) disorders. Here we report the development of a new, minimally invasive behavioral model of abdominal pain induced by
M O Akinwale et al.
African journal of medicine and medical sciences, 41(2), 231-237 (2012-11-29)
The spinal route of analgesia has consolidated its place as a major modality in the management of both acute and chronic pain. The search for ideal additives to local anaesthetic agents to prolong the analgesic effects poses a challenge to
G V Cammu et al.
Anaesthesia and intensive care, 40(6), 999-1006 (2012-12-01)
Six years ago, a study performed in our department reported that the incidence of postoperative residual curarisation (PORC) was 39%. The reassessment of neuromuscular monitoring and reversal of neuromuscular block in routine anaesthetic practice is relevant now that sugammadex has
Comparing sugammadex and neostigmine reversal of neuromuscular blockade in laparoscopic surgery.
D Jain et al.
Anaesthesia, 68(3), 306-307 (2013-02-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service