Skip to Content
Merck
All Photos(1)

Key Documents

IRMM315

4-Deoxynivalenol in acetonitrile

IRMM®, certified reference material

Synonym(s):

Deoxynivalenol solution, 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

application(s)

general analytical

format

matrix material

storage temp.

2-8°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Analysis Note

For more information please see:
IRMM315

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia M Cano et al.
PloS one, 8(1), e53647-e53647 (2013-01-18)
Deoxynivalenol (DON) is a mycotoxin produced by Fusarium species which is commonly found in temperate regions worldwide as a natural contaminant of cereals. It is of great concern not only in terms of economic losses but also in terms of
Joelma Lucioli et al.
Toxicon : official journal of the International Society on Toxinology, 66, 31-36 (2013-02-14)
Trichothecenes induce changes in the intestinal barrier function through decreased expression of cell junction proteins and apoptosis of enterocytes. The mitogen activated protein kinases (MAPK) play an important role in the signaling pathways of cell turnover and differentiation. Using ex
Dirk Rohweder et al.
Archives of animal nutrition, 67(1), 37-47 (2013-01-23)
Fusarium infections do not only affect the grain, but also the rest of the plant, which result in contamination of plants with the mycotoxin deoxynivalenol (DON). The bioavailability of DON may be influenced by the matrix due to the differences
Amal Halawa et al.
Archives of animal nutrition, 67(2), 134-146 (2013-03-26)
Deoxynivalenol (DON) is one of the most important trichothecenes, due to its worldwide distribution and common contamination of animal feed. It mainly affects the gastrointestinal tract and the immune system with a high susceptibility for swine. Lipopolysaccharides (LPS) are endotoxins
Sven Dänicke et al.
Toxicology letters, 220(2), 172-180 (2013-04-23)
The systemic effects of the Fusarium toxin deoxynivalenol (DON) and of bacterial lipopolysaccharides (LPS) were studied in male castrated pigs (40.4 ± 3.7 kg) infused intravenously with either DON or LPS alone (100 μg DON/kg/h, 7.5 μg/LPS/kg/h), or together (100

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service