Skip to Content
Merck
All Photos(1)

Key Documents

C6035

Supelco

Caffeine solution

analytical standard, 1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Caffeine solution may be used as a surrogate standard for the determination of selected antibiotics like ofloxacin, norfloxacin, roxithromycin, erythromycin-H2O, sulfadiazine, sulfadimidine, sulfamethoxazole, amoxicillin and chloramphenicol in water samples using high performance liquid chromatography coupled with electrospray ionization tandem mass spectrometry(HPLC-MS-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of selected antibiotics in the Victoria Harbour and the Pearl River, South China using high-performance liquid chromatography-electrospray ionization tandem mass spectrometry.
Xu W H, et al.
Environmental Pollution (Barking, Essex : 1987), 145(3), 145(3)-145(3) (2007)
Nancy N Maserejian et al.
American journal of epidemiology, 177(12), 1399-1410 (2013-06-01)
Evidence to substantiate recommendations for restriction of caffeinated or acidic beverages as self-management for lower urinary tract symptoms (LUTS) is limited. We examined longitudinal and acute associations between beverage intake and LUTS in the Boston Area Community Health (BACH) cohort
Monica L Bertoia et al.
The American journal of clinical nutrition, 97(6), 1356-1363 (2013-04-26)
Alcohol and caffeine intakes may play a role in the development of sudden cardiac death (SCD) because of their effects on cholesterol, blood pressure, heart rate variability, and inflammation. Our objective was to examine the association between long-term alcohol and
Biomedicine. Human stem cells from cloning, finally.
Gretchen Vogel
Science (New York, N.Y.), 340(6134), 795-795 (2013-05-21)
Rachel A Newman et al.
Perceptual and motor skills, 116(1), 280-293 (2013-07-09)
Naps are an effective strategy for maintaining alertness and cognitive performance; however, upon abrupt wakening from naps, sleep inertia (temporary performance degradation) may ensue. In the present study, attenuation of post-nap sleep inertia was attempted by administration of caffeine gum.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service