Skip to Content
Merck
All Photos(1)

Documents

C2250000

Clindamycin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33ClN2O5S · HCl
CAS Number:
Molecular Weight:
461.44
Beilstein:
4070786
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clindamycin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)C2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H33ClN2O5S.ClH/c1-5-6-10-7-11(21(3)8-10)17(25)20-12(9(2)19)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,22-24H,5-8H2,1-4H3,(H,20,25);1H/t9-,10+,11-,12+,13-,14+,15+,16+,18+;/m0./s1

InChI key

AUODDLQVRAJAJM-XJQDNNTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clindamycin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Clindamycin is a semi-synthetic, lincosamide antibiotic that is prepared from lincomycin. It inhibits bacterial protein synthesis by hydrogen bond interactions with the 23S rRNA component of the 50S ribosomal subunit thus inducing dissociation of the peptidyl-t-RNA complex. It has antibacterial activity against Gram-positive cocci and antiprotozoal activity against Taxoplasma.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Antibacterial and antiprotozoal antibiotic of the lincosamide class.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Lact. - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John W Barlow et al.
BMC veterinary research, 9, 28-28 (2013-02-13)
Treatment of subclinical mastitis during lactation can have both direct (individual animal level) and indirect (population level) effects. With a few exceptions, prior research has focused on evaluating the direct effects of mastitis treatment, and to date no controlled field
D Gonzalez et al.
Clinical pharmacology and therapeutics, 96(4), 429-437 (2014-06-21)
Clindamycin is commonly prescribed to treat children with skin and skin-structure infections (including those caused by community-acquired methicillin-resistant Staphylococcus aureus (CA-MRSA)), yet little is known about its pharmacokinetics (PK) across pediatric age groups. A population PK analysis was performed in
Ronald N Jones et al.
Diagnostic microbiology and infectious disease, 75(3), 304-307 (2013-01-30)
Dalbavancin (DAL) is an investigational lipoglycopeptide with a prolonged serum half-life allowing once weekly dosing. DAL potency was assessed in the 2011 SENTRY Antimicrobial Surveillance Program among 1555 isolates sampled from all 9 US Census regions. Monitored Gram-positive cocci included
Katrin Schilcher et al.
The Journal of infectious diseases, 210(3), 473-482 (2014-02-15)
The Gram-positive human pathogen Staphylococcus aureus causes a variety of human diseases such as skin infections, pneumonia, and endocarditis. The micrococcal nuclease Nuc1 is one of the major S. aureus virulence factors and allows the bacterium to avoid neutrophil extracellular
Michael C Jewett et al.
Molecular systems biology, 9, 678-678 (2013-06-27)
Purely in vitro ribosome synthesis could provide a critical step towards unraveling the systems biology of ribosome biogenesis, constructing minimal cells from defined components, and engineering ribosomes with new functions. Here, as an initial step towards this goal, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service