Skip to Content
Merck
All Photos(3)

Key Documents

B34656

Sigma-Aldrich

Biphenyl

ReagentPlus®, 99.5%

Synonym(s):

1,1′-Biphenyl, Diphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
Beilstein:
1634058
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

5.31 (vs air)

Quality Level

vapor pressure

9.46 mmHg ( 115 °C)

product line

ReagentPlus®

Assay

99.5%

form

flakes
powder or crystals

autoignition temp.

1004 °F

expl. lim.

0.6 %, 111 °F
5.8 %, 166 °F

bp

255 °C (lit.)

mp

68-70 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Biphenyl (BP) is a p-polyphenyl containing two phenyl rings with a non-planar conformation. Its synthesis from benzene by intermolecular direct C-H homocoupling in the presence of palladium-containing catalytic system has been described.[1] Stereochemical studies of its structure have been reported.[2] The nitration of BP using 95% nitric acid to form mononitrobiphenyl isomers has been investigated.[3]

Application

  • Application of Sublimation in the Synthesis and Crystal Growth of Organosulfones: Utilizing sublimation techniques for the synthesis and growth of organosulfone crystals, this study demonstrates how biphenyl derivatives can be integrated into advanced material science applications (Refat et al., 2024).
  • Bicarbazole-Benzophenone Based Twisted Donor-Acceptor Derivatives: Research on bicarbazole-benzophenone derivatives highlights their role as potential blue thermally activated delayed fluorescence emitters for OLED applications, showcasing the utility of biphenyl frameworks in the development of electronic materials (Siddiqui et al., 2024).
  • Optimization of Gas Chromatography Methods for Organic Contaminants: A study details the optimization of a gas chromatography method coupled with Orbitrap mass spectrometry, emphasizing the detection of biphenyl-based organic contaminants in biological samples, crucial for environmental monitoring (Oró-Nolla et al., 2024).
  • Enhanced Solubility and Bioavailability of Antifungal Agents: This investigation presents how biphenyl structures can improve the solubility and bioavailability of clotrimazole, proposing enhanced antifungal treatments through the use of hydrophobized hyperbranched polyglycidol (Jaworska-Krych et al., 2024).
  • Bioremediation of Persistent Organic Pollutants: A systematic review discusses the use of microalgae-bacteria consortia for the bioremediation of biphenyl-containing persistent organic pollutants, providing a sustainable approach to environmental cleanup (Guo et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elucidation of the Forces Governing the Stereochemistry of Biphenyl.
Jia J, et al.
European Journal of Organic Chemistry, 2013(3), 611-616 (2013)
Lujiang Xu et al.
Waste management (New York, N.Y.), 92, 97-106 (2019-06-05)
Terephthalonitrile (TPN) was directly produced from polyethylene terephthalate (PET) plastic via catalytic fast pyrolysis with ammonia. The optimal condition for producing TPN was over 1 g γ-Al2O3-2 wt% catalyst at 500 °C under carrier gas (50% NH3 and 50% N2) with yield of
Zeolite-assisted nitration of biphenyl using nitric acid.
Tai Y, et al.
Research on Chemical Intermediates, 41(2), 867-872 (2015)
Highly Active Palladium-Based Catalyst System for the Aerobic Oxidative Direct Coupling of Benzene to Biphenyl.
Liu Y, et al.
ChemCatChem, 8(2), 448-454 (2016)
Sheng-Hao Hsu et al.
Acta biomaterialia, 8(11), 4151-4161 (2012-07-31)
Low-shrinkage resin-based photocurable liquid crystalline epoxy nanocomposite has been investigated with regard to its application as a dental restoration material. The nanocomposite consists of an organic matrix and an inorganic reinforcing filler. The organic matrix is made of liquid crystalline

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service