Skip to Content
Merck
All Photos(1)

Key Documents

A2250

Supelco

Warfarin

analytical standard

Synonym(s):

4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, Coumafene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
Beilstein:
8868198
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

162-164 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

CC(=O)CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3

InChI key

PJVWKTKQMONHTI-UHFFFAOYSA-N

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Repr. 1A - STOT RE 1 Oral

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Yoshioka et al.
Journal of artificial organs : the official journal of the Japanese Society for Artificial Organs, 17(4), 308-314 (2014-07-23)
The aim of this study was to evaluate our clinical experience with the Jarvik 2000 axial flow pump (Jarvik Heart, Inc, New York, NY, USA), a miniature axial flow left ventricular assist device (LVAD). The clinical results of eight patients
Periprocedural management of new oral anticoagulants in patients undergoing atrial fibrillation ablation.
Jeffrey I Weitz et al.
Circulation, 129(16), 1688-1694 (2014-04-23)
Shinji Kamisuki et al.
Journal of medicinal chemistry, 54(13), 4923-4927 (2011-05-13)
Fatostatin, a recently discovered small molecule that inhibits activation of sterol regulatory element-binding protein (SREBP), blocks biosynthesis and accumulation of fat in obese mice. We synthesized and evaluated a series of fatostatin derivatives. Our structure-activity relationships led to the identification
Tomasz Baron et al.
Heart (British Cardiac Society), 101(2), 101-106 (2014-10-22)
We aimed to assess differences in incidence, clinical features, current treatment strategies and outcome in patients with type 2 vs. type 1 acute myocardial infarction (AMI). All 20 138 hospitalisations in Sweden with a diagnosis of AMI registered during 2011 in
Minoru Yabuta et al.
Journal of vascular and interventional radiology : JVIR, 25(9), 1406-1412 (2014-05-24)
To evaluate retrospectively the long-term outcomes of percutaneous transhepatic balloon angioplasty performed for portal vein stenosis (PVS) after pediatric living donor liver transplantation (LDLT). Between October 1997 and December 2013, of 527 pediatric patients (age < 18 y) who underwent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service