Skip to Content
Merck
All Photos(1)

Key Documents

A0100000

Acetazolamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acetazolamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Nc1nnc(s1)S(N)(=O)=O

InChI

1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)

InChI key

BZKPWHYZMXOIDC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbonic anhydrase inhibitor; increases cerebral blood flow.

Biochem/physiol Actions

Inhibits water permeability of membranes by interacting with aquaporins

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karine Auré et al.
Neurology, 81(21), 1810-1818 (2013-10-25)
To report that homoplasmic deleterious mutations in the mitochondrial DNA MT-ATP6/8 genes may be responsible for acute episodes of limb weakness mimicking periodic paralysis due to channelopathies and dramatically responding to acetazolamide. Mitochondrial DNA sequencing and restriction PCR, oxidative phosphorylation
Tsogyal D Latshang et al.
JAMA, 308(22), 2390-2398 (2012-12-13)
Many patients with obstructive sleep apnea syndrome (OSA) living near sea level travel to altitude, but this may expose them to hypoxemia and exacerbation of sleep apnea. The treatment in this setting is not established. To evaluate whether acetazolamide and
W G Reiss et al.
The Annals of pharmacotherapy, 30(5), 514-519 (1996-05-01)
To summarize the pharmacology, pharmacokinetics, efficacy, and safety of acetazolamide and to evaluate its therapeutic role in patients with epilepsy. A computerized search of the MEDLINE (OVID) database (1966-1994) was used to identify publications regarding acetazolamide. The MEDLINE search was
Paolo Salvi et al.
Hypertension (Dallas, Tex. : 1979), 61(4), 793-799 (2013-02-27)
High-altitude tourism is increasingly frequent, involving also subjects with manifest or subclinical coronary artery disease. Little is known, however, on the effects of altitude exposure on factors affecting coronary perfusion. The aim of our study was to assess myocardial oxygen
Özlen Güzel-Akdemir et al.
Bioorganic & medicinal chemistry, 21(6), 1386-1391 (2013-01-29)
By using phthalimido-substituted aromatic sufonamides as lead molecules, a series of new sulfonamides incorporating ortho-benzenedisulfonimide moieties have been synthesized and tested against the human (h) cytosolic carbonic anhydrase (CA, EC 4.2.1.1) isozymes hCA I and hCA II and the transmembrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service