Skip to Content
Merck
All Photos(1)

Key Documents

108146

Sigma-Aldrich

Benzenesulfonamide

≥98%

Synonym(s):

Phenylsulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2NH2
CAS Number:
Molecular Weight:
157.19
Beilstein:
1100566
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39093209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

form

solid

mp

149-152 °C (lit.)

solubility

methanol: soluble 25 mg/mL

SMILES string

NS(=O)(=O)c1ccccc1

InChI

1S/C6H7NO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H2,7,8,9)

InChI key

KHBQMWCZKVMBLN-UHFFFAOYSA-N

Gene Information

human ... CA1(759)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzenesulfonamide was used to develop analytical method for simultaneous determination of benzotriazole, benzothiazole and benzenesulfonamide contaminants in environmental waters[1].

Biochem/physiol Actions

Benzenesulfonamide is an inhibitor of human carbonic anhydrase B[2]. Benzenesulfonamide derivatives are effective in the treatment of proliferative diseases such as cancer[3]. It is used in the synthesis of dyes, photochemicals and disinfectants[1].

Preparation Note

Benzenesulfonamide dissolves in methanol at a concentration of 25 mg/ml to form a clear, colourless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theres Ramenda et al.
Amino acids, 44(4), 1167-1180 (2013-01-12)
Cu(I)-mediated [3+2]cycloaddition between azides and alkynes has evolved into a valuable bioconjugation tool in radiopharmaceutical chemistry. We have developed a simple, convenient and reliable radiosynthesis of 4-[18F]fluoro-N-methyl-N-(propyl-2-yn-1-yl)benzenesulfonamide ([18F]F-SA) as a novel aromatic sulfonamide-based click chemistry building block. [18F]F-SA could be
Nitrogen-15 nuclear magnetic resonance study of benzenesulfonamide and cyanate binding to carbonic anhydrase.
K Kanamori et al.
Biochemistry, 22(11), 2658-2664 (1983-05-24)
Yousuke Takaoka et al.
Chemical communications (Cambridge, England), 49(27), 2801-2803 (2013-02-27)
Here we describe how a (19)F-probe incorporated into an endogenous protein by a chemical biology method revealed protein dynamics. By explicit determination of ligand-bound and unbound structures with X-ray crystallography, the quantitative comparison of the protein's dynamics in live cells
Genliang Lu et al.
Journal of medicinal chemistry, 56(2), 510-520 (2012-12-14)
Carbonic anhydrase IX (CA-IX) is upregulated in cancer in response to the hypoxic tumor microenvironment, making it an attractive molecular target for the detection of hypoxic solid tumors. A series of small molecule benzenesulfonamide based CA-IX inhibitors containing novel tridentate
Stefano Costanzi et al.
Bioorganic & medicinal chemistry, 20(17), 5254-5261 (2012-07-27)
The P2Y(1) receptor (P2Y(1)R) is a G protein-coupled receptor naturally activated by extracellular ADP. Its stimulation is an essential requirement of ADP-induced platelet aggregation, thus making antagonists highly sought compounds for the development of antithrombotic agents. Here, through a virtual

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service