92107
O-Acetyl-L-carnitine hydrochloride
analytical standard
Synonym(s):
(R)-3-Acetoxy-4-(trimethylammonio)butyrate hydrochloride, ALC, ALCAR, R-(−)-2-Acetyloxy-3-carboxy-N,N,N-trimethyl-1-propanaminium chloride
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥96.0% (HPLC)
96.0-104.0% (AT)
form
powder or crystals
optical activity
[α]/D -27.0±2.0°, c = 1 in H2O
shelf life
limited shelf life, expiry date on the label
color
white to off-white
application(s)
clinical testing
format
neat
storage temp.
2-8°C
SMILES string
C[N+](C)(C)C[C@H](OC(C)=O)CC(O)=O.[Cl-]
InChI
1S/C9H17NO4.ClH/c1-7(11)14-8(5-9(12)13)6-10(2,3)4;/h8H,5-6H2,1-4H3;1H/t8-;/m1./s1
InChI key
JATPLOXBFFRHDN-DDWIOCJRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service