Skip to Content
Merck
All Photos(1)

Key Documents

90893

Supelco

Butylamine

analytical standard

Synonym(s):

1-Aminobutane, n-Butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605269
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.5 (vs air)

vapor pressure

68 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

594 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.401 (lit.)

bp

78 °C (lit.)

mp

−49 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCN

InChI

1S/C4H11N/c1-2-3-4-5/h2-5H2,1H3

InChI key

HQABUPZFAYXKJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Butylamine may be used as a reference standard for the determination of the analyte in sewage water using reversed-phase high-performance liquid chromatography with electrochemical detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of sewage organic chlorination products that are resistant to dechlorination with sulfite.
MacCrehan AW, et al.
Environmental Science & Technology, 32(22), 3640-3645 (1998)
Zachariah M Norman et al.
ACS nano, 8(7), 7513-7521 (2014-06-25)
We report the evolution of electrical transport and grain size during the sintering of thin films spin-cast from soluble phosphine and amine-bound, chloride-terminated cadmium selenide nanocrystals. Sintering of the nanocrystals occurs in three distinct stages as the annealing temperature is
Tomáš Taubner et al.
International journal of biological macromolecules, 72, 11-18 (2014-08-12)
Carboxymethylcellulose (CMC) was selected as substrate for amidation based on previous results described for monocarboxy cellulose (MCC) with the aim to prepare highly substituted products. In comparison with MCC containing uronic carboxyl groups at C-6 position, O-carboxymethyl groups in CMC
Robert Jirásko et al.
Analytical and bioanalytical chemistry, 406(19), 4601-4610 (2014-05-21)
The combination of ultrahigh-resolution mass spectrometry imaging (UHRMSI) and ultrahigh-performance liquid chromatography coupled with tandem mass spectrometry (UHPLC/MS/MS) was used for the identification and the spatial localization of atorvastatin (AT) and its metabolites in rat tissues. Ultrahigh-resolution and high mass
István Ilisz et al.
Journal of chromatography. A, 1384, 67-75 (2015-02-11)
The enantiomers of four unusual, rather rigid isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on a quinine- or a quinidine-based zwitterionic ion-exchanger as chiral selector. The effects of the mobile phase composition, the structures of the analytes and temperature on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service