Skip to Content
Merck
All Photos(2)

Documents

901767

Sigma-Aldrich

PC biotin-PEG3-NHS ester

Synonym(s):

PC biotin-NHS ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H52N6O15S
CAS Number:
Molecular Weight:
840.89
UNSPSC Code:
51171641
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

mp

170 °C

storage temp.

−20°C

Application

PC biotin-NHS ester is a unique amine-reactive, photocleavable biotin reagent that allows for reagent-free release of the captured biomolecules from streptavidin. This reagent contains a biotin moiety linked to the amine reactive N-hydroxysuccinimidyl (NHS) mixed carbonate through a spacer arm containing a photocleavable moiety. The NHS portion of this compound reacts specifically with primary amine groups on the target molecule(s) to form a carbamate linkage. Captured biomolecules can be efficiently photoreleased, typically >90% in 5-25 minutes using an inexpensive, near-UV, low intensity lamp (e.g. 365 nm lamp at 1-5 mW/cm2).

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service