Skip to Content
Merck
All Photos(2)

Documents

423564

Sigma-Aldrich

4-Bromomethyl-3-nitrobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
260.04
Beilstein:
1970939
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

127-130 °C (lit.)

solubility

DMF: soluble(lit.)
dichloromethane: soluble(lit.)

SMILES string

OC(=O)c1ccc(CBr)c(c1)[N+]([O-])=O

InChI

1S/C8H6BrNO4/c9-4-6-2-1-5(8(11)12)3-7(6)10(13)14/h1-3H,4H2,(H,11,12)

InChI key

QMAHVAFURJBOFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromomethyl-3-nitrobenzoic acid (BNBA) is a benzoic acid derivative. It has been synthesized by the nitration of 4-bromomethylbenzoic acid using fuming nitric acid. It participates in the synthesis of 3,4-dihydro-2(1H)-quinazolinones and 3,4-dihydro-1H-quinazolin-2-thiones.

Application

4-Bromomethyl-3-nitrobenzoic acid may be used in the following studies:
  • As a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester (BNBA-SE).
  • As a reactant in the synthesis of 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid.
  • As a reactant in the synthesis of decyl 4-(bromomethyl)-3-nitrobenzoate.
  • As a reactant in the synthesis of 4-((2-(hydroxymethyl)phenylamino)methyl)-3-nitrobenzoic acid.
  • As a thiol photo-deprotection reagent.
  • As a UV-cleavable reagent to functionalize polyacrylamide acryl hydrate (PAAH).
  • As a photocleavable linker to form localized photoinversion of surface charges inside the colloidal crystal.
  • As a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.
  • As a reactant in the synthesis of (N-allyloxycarbonyl)-5-aminopent-1-yl 4-bromomethyl-3-nitrobenzoate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase synthesis of 3, 4-dihydro-2 (1H)-quinazolinones and 3, 4-dihydro-1H-quinazolin-2-thiones.
Sun Q, et al.
Tetrahedron Letters, 42(25), 4119-4121 (2001)
Hai-Yuan Hsu et al.
Molecular diversity, 16(2), 241-249 (2011-12-20)
An efficient microwave-assisted and water-soluble ionic liquid (IL)-supported synthesis of medicinally important dihydro- and tetrahydroquinazolines has been developed. The protocol involves the S(N)2 substitution reaction of IL-bound 4-bromomethyl-3-nitrobenzoic acid with various primary amines to provide IL-bound 4-((alkylamino) methyl)-3-nitrobenzoate under microwave
Eric Besson et al.
Langmuir : the ACS journal of surfaces and colloids, 22(20), 8346-8352 (2006-09-20)
This work describes how selective patterning of hydrophobic and hydrophilic areas inside microchannels of microfluidic devices can be achieved by combining well-known chemical protocols and standard photolithography equipment (365 nm). Two techniques have been performed and compared. The first technique
Margo T Frey et al.
Soft matter, 5, 1918-1924 (2009-08-13)
Recent studies indicate that extracellular mechanical properties, including rigidity, profoundly affect cellular morphology, growth, migration, and differentiation [R. J. Pelham, Jr. and Y. Wang, Proc. Natl. Acad. Sci. U. S. A., 1997, 94(25), 13661-13665; H. B. Wang, M. Dembo and
G Marriott et al.
Biochemistry international, 26(5), 943-951 (1992-04-11)
The ability to generate substrate concentration jumps through photo-deprotection of amine, carboxyl and phosphate groups has been an important development for investigations of protein activity in complex systems. To broaden the versatility and applications of photo-deprotection techniques for the photomodulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service