900519
Isopropyl acetate
anhydrous, ZerO2®, ≥99.6%
Synonym(s):
Degassed and low oxygen isopropyl acetate, IPrOAc, Isopropyl acetate ZerO2®, Acetic acid isopropyl ester
About This Item
Recommended Products
grade
ZerO2®
Quality Level
vapor density
3.5 (vs air)
vapor pressure
47 mmHg ( 20 °C)
Assay
≥99.6%
form
liquid
autoignition temp.
894 °F
quality
anhydrous
expl. lim.
1.8 %, 37 °F
8 %
greener alternative product characteristics
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
impurities
≤0.1% water
water
refractive index
n20/D 1.377 (lit.)
bp
85-91 °C (lit.)
mp
−73 °C (lit.)
density
0.872 g/mL at 25 °C (lit.)
greener alternative category
, Aligned
SMILES string
CC(C)OC(C)=O
InChI
1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3
InChI key
JMMWKPVZQRWMSS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
41.0 °F - closed cup
Flash Point(C)
5 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service