89151
Dimethylmalonic acid
Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
About This Item
Recommended Products
grade
Standard for quantitative NMR
certified reference material
TraceCERT®
Quality Level
description
qNMR Standard for D2O and organic solvents (1.3 - 1.4 ppm)
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
technique(s)
gas chromatography (GC): suitable
qNMR: suitable
mp
191-193 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
CC(C)(C(O)=O)C(O)=O
InChI
1S/C5H8O4/c1-5(2,3(6)7)4(8)9/h1-2H3,(H,6,7)(H,8,9)
InChI key
OREAFAJWWJHCOT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Check out our entire range of quantitative NMR standards (qNMR standards)
Application
- Metabolic enzyme research: A study involving ECHDC1 knockout mice, which affect metabolism of branched-chain fatty acids, referenced the role of derivatives like dimethylmalonic acid in understanding metabolic disorders. This research is significant for biochemical studies related to metabolism and enzyme function (Dewulf et al., 2021).
- NMR-based metabolomics: Utilizing NMR techniques, researchers have analyzed metabolites in biological samples, demonstrating the utility of dimethylmalonic acid in enhancing the resolution of metabolic profiling. This methodology is valuable in biochemical research, especially in studying complex biological matrices (Tang & Hatzakis, 2020).
Other Notes
suitable NMR solvents: MeOD, D2O, DMSO-d6
Recommended products
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The signal ratio of two different protons can be measured with tremendous precision, which enables the generation of certified reference materials for use as qNMR standards.
NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.
NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.
NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.
Related Content
ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.
ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.
ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service