Skip to Content
Merck
All Photos(3)

Documents

857343

Sigma-Aldrich

Basic Fuchsin

certified by the Biological Stain Commission, Dye content ≥88 %

Synonym(s):

Basic Parafuchsin, Basic Red 9, Magenta O, Parafuchsin hydrochloride, Paramagenta hydrochloride, Pararosaniline chloride, Pararosaniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3 · HCl
CAS Number:
Molecular Weight:
323.82
Colour Index Number:
42500
Beilstein:
4164603
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥88%

color

green to dark green

pH

1.0-3.1, purple to red

mp

268-270 °C (dec.) (lit.)

λmax

544 nm

ε (extinction coefficient)

≥11000 at 235-239 nm in 50% ethanol at 0.003 g/L
≥17000 at 287-291 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Nc1ccc(cc1)C(\c2ccc(N)cc2)=C3/C=CC(=[NH2+])C=C3

InChI

1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H

InChI key

JUQPZRLQQYSMEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Basic fuchsin has been used as a lignin-detecting fluorogenic dye for fluorescence-based imaging of lignin in the basal internodes of three Brachypodium distachyon ecotypes.

Biochem/physiol Actions

Basic fuchsin belongs to the triphenylmethane group. Combining basic fuchsin with sulfurous acid results in the formation of fuchsin sulfurous acid, also referred to as Feulgen reagent or Schiff′s reagent. This reagent is responsible for the staining of chromosomal DNA. Basic fuchsin is also used for the staining of lignin in plant tissue samples.

Suitability

Certified for use for acid-fast staining with carbol-fuchsin, as ENDO medium, and as Schiff′s reagent in the Periodic Acid Schiff (PAS) and Feulgen techniques.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bisen PS
Laboratory Protocols in Applied Life Sciences (2014)
Min Chen et al.
The New phytologist, 229(6), 3481-3496 (2020-11-25)
The intimate association of host and fungus in arbuscular mycorrhizal (AM) symbiosis can potentially trigger induction of host defence mechanisms against the fungus, implying that successful symbiosis requires suppression of defence. We addressed this phenomenon by using AM-defective vapyrin (vpy)
Plant Fibers (2012)
Sharma A
Plant Chromosomes (1999)
M E Baker et al.
The Analyst, 119(5), 959-961 (1994-05-01)
The colorimetric indicator pararosaniline has been immobilized onto the cation-exchange resins Amberlite IRC-50, Dowex 50W-X8 and cellulose phosphate by electrostatic bonding. The reflectance of each reagent phase was measured using a bifurcated fibre-optic system and a flow cell. Pararosaniline immobilized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service