Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Basic Fuchsin
Synonym(s):
Basic Parafuchsin, Basic Red 9, Magenta
™
O, Parafuchsin hydrochloride, Paramagenta hydrochloride, Pararosaniline chloride, Pararosaniline hydrochloride
Empirical Formula (Hill Notation):
C
19
H
17
N
3
· HCl
CAS Number:
569-61-9
Molecular Weight:
323.82
EC Number:
209-321-2
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
215597
Basic Fuchsin,
Dye content >85 %
Expand
View Pricing
857343
Basic Fuchsin,
certified by the Biological Stain Commission, Dye content ≥88 %
Expand
View Pricing