76244
Paraffin wax
pellets, white, tested according to Ph. Eur.
Synonym(s):
Paraffinum solidum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
USP/NF
tested according to Ph. Eur.
Quality Level
form
pellets
color
white
mp
50-61 °C
application(s)
pharmaceutical (small molecule)
InChI key
JWHAUXFOSRPERK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
602.6 °F - Pensky-Martens closed cup
Flash Point(C)
317 °C - Pensky-Martens closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Archives of physical medicine and rehabilitation, 94(4), 642-649 (2012-11-29)
To evaluate the efficacy of paraffin bath therapy on pain, function, and muscle strength in patients with hand osteoarthritis. Prospective single-blinded randomized controlled trial. Department of physical medicine and rehabilitation in a university hospital. Patients with bilateral hand osteoarthritis (N=56).
Journal of hazardous materials, 243, 105-111 (2012-10-31)
In this study, nanoscale zero-valent iron (NZVI) particles were synthesized and used for the reductive dehalogenation of short chain chlorinated paraffins (SCCPs) in the laboratory. The results show that the dechlorination rate of chlorinated n-decane (CP(10)) by NZVI increased with
Journal of chromatography. A, 1274, 36-43 (2013-01-05)
Accurately quantifying short chain chlorinated paraffins (SCCPs) in soil samples with gas chromatograph coupled with electron capture negative ionization mass spectrometry (GC-ECNI-MS) is difficult because many other polychlorinated pollutants are present in the sample matrices. These pollutants (e.g., polychlorinated biphenyls
[Paraffin throughout history].
Annales de dermatologie et de venereologie, 140(2), 154-156 (2013-02-12)
Diagnostic molecular pathology : the American journal of surgical pathology, part B, 22(1), 52-58 (2013-02-02)
We report a hydrothermal pressure method (pressure cooking) for simultaneous deparaffinization and lysis of formalin-fixed paraffin-embedded tissue followed by conventional chaotropic salt column purification to obtain high-quality DNA. Using this method, the release of DNA occurred within the first minute
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service