Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Paraffin wax
CAS Number:
8002-74-2
Products
Product Category
lipids (5)
Brand
Sigma-Aldrich (9)
Color
Feature
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Special Grade
Purum (2)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
327204
Paraffin wax,
mp 53-58 °C (ASTM D 87)
Expand
View Pricing
327212
Paraffin wax,
mp 58-62 °C (ASTM D 87)
Expand
View Pricing
76242
Paraffin wax,
pellets, white
Expand
View Pricing
411663
Paraffin wax,
mp ≥65 °C (ASTM D 87)
Expand
View Pricing
76228
Paraffin wax,
chunks (large), white
Expand
View Pricing
18634
Paraffin wax,
meets analytical specification of Ph. Eur., pastilles, white
Expand
View Pricing
76244
Paraffin wax,
pellets, white, tested according to Ph. Eur.
Expand
View Pricing
76233
Paraffin wax,
tested according to Ph. Eur., low viscosity
Expand
View Pricing
76234
Paraffin wax,
tested according to Ph. Eur., high viscosity
Expand
View Pricing