Skip to Content
Merck
All Photos(1)

Key Documents

75544

Supelco

1-Dodecanol

analytical standard

Synonym(s):

Alcohol C12, Dodecyl alcohol, Lauryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
Beilstein:
1738860
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

7.4 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

500 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.442 (lit.)

bp

260-262 °C (lit.)

mp

22-26 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCO

InChI

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Dodecanol belongs to a broad range of flavor and fragrance standards, used for the quality control of food and cosmetic products. It is identified as the main component in Etlingera elatior flower, widely used in traditional medicine and as a flavoring compound in food products, essential oils of Hypericum perforatum and Polygonum minus. It is the most studied C12 alcohol and finds extensive usage as a diluent/solvent/blender in low-cost perfumes, detergent fragrances, etc.

Application

1-Dodecanol may be used as an analytical reference standard for the quantification of the analyte in:
  • Etlingera elatior

  • flower using gas chromatography-mass spectroscopy (GC-MS).
  • Essential oils from the aerial parts of Hypericum perforatum by gas chromatography (GC) and gas chromatography coupled to mass spectrometry (GC-MS).
  • Essential oils from Polygonum minus by gas chromatography coupled to mass spectrometry (GC-MS) and GC-MS/olfactometry (GC-MS/O) with aroma extraction dilution analysis (AEDA).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition, phytochemical and antioxidant activity from extract of Etlingera elatior flower from Indonesia
Maimulyanti A and Prihadi RA
Journal of Pharmacognosy and Phytochemistry, 3(6), 233-238 (2015)
GC-MS/Olfactometric characterisation and aroma extraction dilution analysis of aroma active compounds in'Polygonum minus' essential oil
Rusdi AN, et al.
Plant Archives, 9(4), 289-289 (2016)
Changes in essential oil composition in Saint John?s wort (Hypericum perforatum L.) aerial parts during its phenological cycle
Schwob I, et al.
Biochemical Systematics and Ecology, 32(8), 735-745 (2004)
Perfume and Flavor Chemicals (Aroma Chemicals) Vol.1, 1 (2019)
Patty's Toxicology, 6(5) (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service