Skip to Content
Merck
All Photos(1)

Documents

185388

Sigma-Aldrich

1-Tetradecanol

97%

Synonym(s):

Myristyl alcohol, Tetradecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13OH
CAS Number:
Molecular Weight:
214.39
Beilstein:
1742652
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020231
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.4 (vs air)

Quality Level

Assay

97%

form

solid

autoignition temp.

489 °F

bp

289 °C (lit.)

mp

35-39 °C (lit.)

solubility

water: 0.00013 g/L at 23 °C
chloroform: soluble 10%

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCO

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3

InChI key

HLZKNKRTKFSKGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Tetradecanol was used for filling the hollow interiors of gold nanocages in the fabrication of new theranostic system having unique feature of photoacoustic imaging. It was used in fabrication of temperature-regulated drug release system based on phase-change materials.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A temperature-sensitive drug release system based on phase-change materials.
Sung-Wook Choi et al.
Angewandte Chemie (International ed. in English), 49(43), 7904-7908 (2010-09-15)
Geon Dae Moon et al.
Journal of the American Chemical Society, 133(13), 4762-4765 (2011-03-16)
This communication reports a new theranostic system with a combination of capabilities to both enhance the contrast of photoacoustic (PA) imaging and control the release of a chemical or biological effector by high-intensity focused ultrasound (HIFU). The fabrication of this
Antonio Ortiz et al.
Journal of chemical ecology, 30(5), 991-1000 (2004-07-28)
The potential for pheromone-based mating disruption of the olive pyralid moth (OPM), Euzophera pinguis, in olive groves was investigated during the second flight period in small-plot trials in 2002. The female of this species emits a blend of (9Z,12E)-tetradecadien-1-ol and
L Kato et al.
International journal of leprosy and other mycobacterial diseases : official organ of the International Leprosy Association, 62(1), 75-88 (1994-03-01)
Host-grown Mycobacterium leprae cell suspensions oxidized water-soluble complexes of palmitic acid, myristic acid, cetyl alcohol, and myristyl alcohol prepared with randomly methylated-beta-cyclodextrin as host molecules. Gas chromatography analysis showed that the water-soluble complexes retained their chemical structure following sterilization in
M Lafleur et al.
Biochemistry, 29(36), 8325-8333 (1990-09-11)
The orientational order profile has been determined by using deuterium nuclear magnetic resonance (2H NMR) for POPE in the lamellar liquid-crystalline (L alpha) and the hexagonal (HII) phases and is shown to be sensitive to the symmetry of the lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service