Skip to Content
Merck
All Photos(1)

Key Documents

75283

Supelco

N-Nitrosoanabasine

analytical standard

Synonym(s):

1-Nitroso-2-(3-pyridyl)piperidine, 3-(1-Nitroso-2-piperidinyl)pyridine, NAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=NN1CCCCC1c2cccnc2

InChI

1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2

InChI key

BXYPVKMROLGXJI-UHFFFAOYSA-N

General description

N-Nitrosoanabasine is a tobacco specific nitrosamine (TSNAs) and a potent carcinogenic compound, formed as a result of nitrosation of nicotine.

Application

N-Nitrosoanabasine may be used as an analytical reference standard for the quantification of the analyte in snuff samples and cigarettes using different chromatography techniques and standard for Supelco MIP SPE cartridges.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marine Valette et al.
Journal of integrative plant biology, 62(2), 228-246 (2019-03-29)
Plant growth-promoting rhizobacteria (PGPR), whose growth is stimulated by root exudates, are able to improve plant growth and health. Among those, bacteria of the genus Azospirillum were shown to affect root secondary metabolite content in rice and maize, sometimes without
Mary Fisher et al.
Journal of the American Board of Family Medicine : JABFM, 32(5), 663-673 (2019-09-12)
Patient and community engagement is essential to maintain the relevance of practice-based research. Empowered engagement requires going beyond the check box, with advisory groups involved in every aspect of a project. Here, 4 Colorado practice-based research networks (PBRNs) share their
Kelsey R Thomas et al.
Psychiatric rehabilitation journal, 40(1), 70-78 (2016-08-23)
This study explored whether age moderated cognitive, symptom, and functional changes over a 12-week compensatory cognitive training (CCT) intervention for participants with severe mental illnesses. CCT focused on the cognitive domains of attention, learning, prospective memory, and executive functioning, often
Supercritical fluid extraction and gas chromatography/mass spectrometry for the analysis of tobacco-specific nitrosamines in cigarettes
Song S and Ashley.LD
Analytical Chemistry, 71(7), 1303-1308 (1999)
Xiaojing Xia et al.
Drug design, development and therapy, 11, 1417-1427 (2017-05-24)
Oleanolic acid (OA), a pentacyclic triterpene, is used to safely and economically treat hepatopathy. However, OA, a Biopharmaceutics Classification System IV category drug, has low bioavailability owing to low solubility (<1 μg/mL) and biomembrane permeability. We developed a novel OA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service