Skip to Content
Merck
All Photos(1)

Key Documents

75035

Supelco

Mettler-Toledo Calibration substance ME 18872, Caffeine

Mettler-Toledo Calibration substance ME 18872, Caffeine

traceable to primary standards (LGC)

Synonym(s):

Caffeine, 1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

description

for the calibration of the thermosystem 900

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the certificate and label

mp

234-236.5 °C (lit.)

application(s)

cleaning products
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Caffeine Mettler-Toledo melting point standard is useful for the routine calibration of Mettler-Toledo melting point instrument. The value for its melting point equals an average of 6 to 12 measurements with a Mettler-Toledo MP70 excellence melting point instrument that is calibrated with primary standards.

Application

Mettler-Toledo calibration standard ME 18872, caffeine (234-236.5 °C) finds its use in the calibration of thermosystem 900, in both pharmacopeia and thermodynamic modes of operation.
This mp standard is also employed as a temperature calibration standard in thermal analysis techniques.[1][2][3]

Features and Benefits

  • Melting point calibration standard traceable to a primary standard from LGC, London
  • Melting points measured in both thermodynamic and pharmacopeia modes
  • Available with certificates of analysis and safety data sheet
  • A product of analytical standard grade that meets the QC/QA requirements of melting point analysis
  • Ready-to-use melting point standard, supplied as a finely ground powder

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metoprolol: solid forms of a top selling antihypertensive
Ciciliati MA, et al.
CrystEngComm (2019)
Lamotrigine: design and synthesis of new multicomponent solid forms
Evora AO, et al.
European Journal of Pharmaceutical Sciences (2019)
Co-crystals of diflunisal and isomeric pyridinecarboxamides?a thermodynamics and crystal engineering contribution
Evora.L.OA, et al.
CrystEngComm, 18(25), 4749-4759 (2016)
Co-crystals of diflunisal and isomeric pyridinecarboxamides-a thermodynamics and crystal engineering contribution
Evora AO, et al.
CrystEngComm (2016)
Nicolas Bousette et al.
Journal of cellular physiology, 229(3), 374-383 (2013-09-17)
Calnexin (CNX) is an endoplasmic reticulum (ER) quality control chaperone that has been implicated in ER stress. ER stress is a prominent pathological feature of various pathologic conditions, including cardiovascular diseases. However, the role of CNX and ER stress has

Questions

1–2 of 2 Questions  
  1. Are the Mettler-Toledo Calibration melting standards already dried and pulverized, ready to use out of the bottle?

    1 answer
    1. The answer is yes. All melting point standards are ground to a fine powder, tested as melting point standards and ready to use.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service