Skip to Content
Merck
All Photos(1)

Documents

74350

Supelco

2-Picoline

analytical standard

Synonym(s):

α-Picoline, 2-Methylpyridine, NSC 3409

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
Beilstein:
104581
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.2 (vs air)

vapor pressure

10 mmHg ( 24.4 °C)

Assay

≥98.5% (GC)

autoignition temp.

995 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.499-1.502
n20/D 1.500 (lit.)

bp

128-129 °C (lit.)

mp

−70 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1ccccn1

InChI

1S/C6H7N/c1-6-4-2-3-5-7-6/h2-5H,1H3

InChI key

BSKHPKMHTQYZBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Picoline is a flavor and fragrance standard used for the quality control of food and cosmetic products. It is an isomer of methylpyridine and also a volatile flavor compound reportedly identified in dark chocolates, roasted sesame seed oil, wild rice (Zizania palustris L.), etc.

Application

2-Picoline may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Cigarette smoke using reversed-phase high-performance liquid chromatography–electrospray ionization tandem mass spectrometric method (RP-HPLC–ESI-MS/MS).
  • Chinese faint-scent cigarettes using gas chromatography–mass spectrometry (GC–MS) technique.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Review on Volatile Flavor Components of Roasted Oilseeds and Their Products
Cuicui LI, et al.
Green Chemistry, 1(4), 151-156 (2018)
Matrix effects on flavour volatiles release in dark chocolates varying in particle size distribution and fat content using GC?mass spectrometry and GC?olfactometry
Afoakwa OE, et al.
Food Chemistry, 113(1), 208-215 (2009)
Identification of aroma types and their characteristic volatile compounds of Chinese faint-scent cigarettes based on descriptive sensory analysis and GC?MS and partial least squares regression
Yin F, et al.
European Food Research and Technology, 242(6), 869-880 (2016)
Aroma-active compounds of wild rice (Zizania palustris L.)
Cho S and Kays JS
Food Research International, 54(2), 1463-1470 (2013)
Determination of pyridine, 2-picoline, 4-picoline and quinoline from mainstream cigarette smoke by solid-phase extraction liquid chromatography/electrospray ionization tandem mass spectrometry
Saha S, et al.
Journal of Chromatography A, 1217(3), 307-311 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service