Skip to Content
Merck
All Photos(1)

Key Documents

236276

Sigma-Aldrich

3-Methylpyridine

≥99.5%

Synonym(s):

3-Picoline, 3-Methylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
Beilstein:
1366
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

vapor pressure

4.4 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

~1000 °F

refractive index

n20/D 1.504 (lit.)

bp

144 °C (lit.)

mp

−19 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: miscible(lit.)

density

0.957 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1

InChI

1S/C6H7N/c1-6-3-2-4-7-5-6/h2-5H,1H3

InChI key

ITQTTZVARXURQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical oxidation of 3-methylpyridine (3-MP) at synthetic boron-doped diamond (BDD) thin film electrode has been investigated by cyclic voltammetry and bulk electrolysis. 3-Methylpyridine forms adduct with 2,6-dichloro-4-nitrophenol and the structure of adduct has been determined by X-ray diffraction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Li Sun et al.
Journal of reproduction & infertility, 12(3), 215-220 (2011-07-01)
HLA-G is a major histocompatibility complex (MHC), class Ib molecule that is selectively expressed at the fetal-maternal interface. It is thought to play a role in protecting the fetus from the maternal immune response. Interestingly, the preimplantation embryo development (Ped)
Wenzhen Yu et al.
Molecular vision, 19, 1397-1405 (2013-07-05)
Pathological retinal angiogenesis is a major cause of vision loss. Semaphorin 3A (Sema3A), a chemorepellent guidance protein, plays crucial roles in neural and vascular patterning. To identify its role in retinal neovascularization, we investigated its antiangiogenic effects. Human umbilical vein
Marco Panizza et al.
Annali di chimica, 92(10), 995-1006 (2002-12-20)
The electrochemical behaviour of a synthetic boron-doped diamond thin film electrode (BDD) has been studied in acid media containing phenol using cyclic voltammetry and bulk electrolysis. The results have shown that in the potential region of water stability direct electron
Xinyu Yang et al.
Arthritis research & therapy, 17, 140-140 (2015-05-30)
Our previous proteomic study on fibroblast-like synoviocytes (FLSs) derived from the synovial tissues found that the expression of KIAA1199 was higher in rheumatoid arthritis (RA) patients than in healthy controls. The aim of this study was to examine the biological
Structure and spectroscopic behaviour of the solid adduct of 3-methylpyridine with 2, 6-dichloro-4-nitrophenol.
Majerz I, et al.
Journal of Molecular Structure, 297, 177-184 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service