All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.21
form:
pellets
Recommended Products
grade
p.a.
Quality Level
form
pellets
quality
with indicator
loss
13-18% loss on drying, 105 °C
particle size
2-5 mm
pH
7-14
capacity
≥27 % absorption capacity (CO2)
bulk density
~75 g/100 mL
SMILES string
[OH-].[Na+].O=[Ca]
InChI
1S/Ca.Na.H2O.O/h;;1H2;/q;+1;;/p-1
InChI key
HUAUNKAZQWMVFY-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Effect of soda lime on the degradation of sevoflurane has been studied. Degradation products were investigated for their toxicity.
Soda lime is a mixture of calcium hydroxide with sodium or potassium hydroxide. When not stored in sealed containers, it quickly deteriorates due to carbon dioxide and water absorption. In laboratories, it is also used as a drying agent.
Application
Soda lime can be used:
- As a hydrolysis reagent in the synthesis of bentazone from isatoic anhydride.
- In the monitoring of CO2 efflux from field soils.
- In the direct measurement of dissolved CO2 (carbon dioxide) in supersaturated water.
Soda lime may be employed for the conversion of α-amino acids to primary amines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M Morio et al.
Anesthesiology, 77(6), 1155-1164 (1992-12-01)
Sevoflurane previously has been reported to undergo extensive degradation in the presence of soda lime. To more completely characterize the extent and significnce of this reaction, we studied degradation of sevoflurane with and without soda lime, as well as the
T Lombardo et al.
The Science of the total environment, 408(4), 976-984 (2009-11-11)
Several exposure campaigns of silica-soda-lime window glass have been performed in 30 European sites and 1 in Canada in order to understand, quantify and model the phenomenon of soiling. In this purpose samples were exposed sheltered from the rain. Parallel
Chuchai Anunmana et al.
Dental materials : official publication of the Academy of Dental Materials, 25(11), 1453-1458 (2009-08-13)
To test the hypothesis that the indentation crack technique can determine surface residual stresses that are not statistically significantly different from those determined from the analytical procedure using surface cracks, the four-point flexure test, and fracture surface analysis. Soda-lime-silica glass
E C Romani et al.
Optics express, 20(5), 5429-5439 (2012-03-16)
Materials presenting high optical nonlinearity, such as materials containing metal nanoparticles (NPs), can be used in various applications in photonics. This motivated the research presented in this paper, where morphological, linear and nonlinear optical characteristics of gold NPs on the
Farah Mohammadesmaeili et al.
Water environment research : a research publication of the Water Environment Federation, 82(4), 342-350 (2010-05-04)
Lime and soda-ash softening of reclaimed water reverse osmosis concentrates as a pretreatment step for concentration by seawater reverse osmosis was the focus of this study. The objectives were removal of the potential fouling minerals of calcium, magnesium, and silica
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service