Skip to Content
Merck
All Photos(2)

Key Documents

67261

Supelco

Bis(2-ethylhexyl) phthalate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

392.0 °F - Pensky-Martens closed cup

Flash Point(C)

200 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Xu et al.
Indoor air, 25(1), 79-92 (2014-04-29)
Retail stores contain a wide range of products that can emit a variety of indoor pollutants. Among these chemicals, phthalate esters and polybrominated diphenyl ethers (PBDEs) are two important categories of semi-volatile organic compounds (SVOCs). Filters in heating, ventilation, and
Na Rae Song et al.
Environment international, 54, 65-73 (2013-02-19)
Di(2-ethylhexyl) phthalate (DEHP) is one of the common phthalate plasticizers used primarily in soft polyvinyl chloride, which is a plastic polymer that makes up the total weight of goods from 1% up to 40% in many consumer products. The aims
N B Hopf et al.
Toxicology letters, 224(1), 47-53 (2013-10-22)
Phthalates are suspected to be endocrine disruptors. Di(2-ethylhexyl) phthalate (DEHP) is assumed to have low dermal absorption; however, previous in vitro skin permeation studies have shown large permeation differences. Our aims were to determine DEHP permeation parameters and assess extent
D B Martinez-Arguelles et al.
The Journal of steroid biochemistry and molecular biology, 137, 5-17 (2013-01-22)
Di-(2-ethylhexyl) phthalate (DEHP) is a plasticizer with endocrine disrupting properties that is found ubiquitously in the environment as well as in human amniotic fluid, umbilical cord blood, human milk, semen, and saliva. It is used in the industry to add
Anne-Marie Saillenfait et al.
Journal of applied toxicology : JAT, 33(9), 1027-1035 (2013-06-12)
In utero exposure to the phthalate ester plasticizer di-n-hexyl phthalate (DnHP) is known to affect the development of the male reproductive system and induce alterations in androgen-dependent tissues of male rat offspring. Male reproductive malformations produced by several phthalates have

Articles

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

Identification and quantification of the most common extractables originating from single-use equipment by GC/MS and an Extractables and Leachables Screening Standard.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service