Skip to Content
Merck
All Photos(1)

Key Documents

66742

Supelco

Dichloromethane

Selectophore, ≥99.5%

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
grade:
for ion-selective electrodes
Assay:
≥99.5% (GC)
≥99.5%
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

for ion-selective electrodes

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

description

solvent for preparation of membranes

product line

Selectophore

Assay

≥99.5% (GC)
≥99.5%

form

liquid

autoignition temp.

1223 °F

contains

~0.005% amylene as stabilizer

expl. lim.

22 %

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.324-1.326 g/mL at 20 °C
1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application


  • New Two-Photon Absorbing Squaraine Derivative with Efficient Near-Infrared Fluorescence, Superluminescence, and High Photostability.: This research investigates a novel squaraine derivative, noting its application in two-photon absorption. The use of dichloromethane in the solvent system is critical for achieving high photostability and fluorescence properties in near-infrared applications (Bondar et al., 2022).


  • Identification of Antibacterial Sterols from Korean Wild Mushroom Daedaleopsis confragosa via Bioactivity- and LC-MS/MS Profile-Guided Fractionation.: This study utilized dichloromethane for the extraction of antibacterial sterols from the Daedaleopsis confragosa mushroom. The findings contribute to understanding the chemical properties and potential therapeutic uses of natural extracts (Na et al., 2022).


  • Anti-Inflammatory Activity and Chemical Analysis of Different Fractions from Solidago chilensis Inflorescence.: This research highlights the anti-inflammatory properties of Solidago chilensis inflorescence, with an emphasis on the role of dichloromethane in fractionating and analyzing the plant′s active chemical components (de Brito et al., 2021).


  • Evaluation of analytical method for polycyclic aromatic hydrocarbons content in home meal replacement products by GC/MS.: This publication discusses the development of an analytical method for detecting polycyclic aromatic hydrocarbons in food products, where dichloromethane is utilized as part of the extraction process to ensure accuracy and efficiency in the GC/MS analysis (Lee et al., 2021).


Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Golsteijn et al.
Environment international, 71, 36-45 (2014-06-28)
The present paper describes a method that accounts for variation in indoor chemical exposure settings and accompanying human toxicity in life cycle assessment (LCA). Metal degreasing with dichloromethane was used as a case study to show method in practice. We
Shinji Kumagai et al.
Occupational and environmental medicine, 70(7), 508-510 (2013-03-16)
The present study was conducted to investigate the relationship between occupational chemical exposure and incidence of cholangiocarcinoma among workers in the offset colour proof-printing section of a small printing company in Osaka, Japan. We identified 51 men who had worked
K E Mauck et al.
Integrative and comparative biology, 54(2), 193-209 (2014-04-22)
Recent research suggests that plant viruses, and other pathogens, frequently alter host-plant phenotypes in ways that facilitate transmission by arthropod vectors. However, many viruses infect multiple hosts, raising questions about whether these pathogens are capable of inducing transmission-facilitating phenotypes in
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
Guilherme Asmar Alencar et al.
The Journal of clinical endocrinology and metabolism, 99(8), E1501-E1509 (2014-04-09)
Primary macronodular adrenal hyperplasia (PMAH) is a rare cause of Cushing's syndrome, usually characterized by functioning adrenal macronodules and increased cortisol production. Familial clustering of PMAH has been described, suggesting an inherited genetic cause for this condition. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service