Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dichloromethane
Synonym(s):
Methylene chloride
Empirical Formula (Hill Notation):
CH
2
Cl
2
CAS Number:
75-09-2
Molecular Weight:
84.93
EC Number:
200-838-9
Products
Product Category
solvents (23)
coatings (1)
pesticides (1)
Brand
SAFC (1)
Sigma-Aldrich (14)
Supelco (11)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (5)
Puriss (4)
Anhydrous (3)
Puriss P.A. (2)
Biotechnology (1)
analytical standard (1)
Technique
Filter Products
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
34856
Dichloromethane,
suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Expand
View Pricing
D65100
Dichloromethane,
contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Expand
View Pricing
650463
Dichloromethane,
HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Expand
View Pricing
270997
Dichloromethane,
anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Expand
View Pricing
1.06050
Dichloromethane,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
320269
Dichloromethane,
ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Expand
View Pricing
1.06044
Dichloromethane,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
5.89581
Dichloromethane,
99.8%, anhydrous, contains 40-150 ppm amylene as stabilizer
Expand
View Pricing
32222
Dichloromethane,
puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Expand
View Pricing
32222-M
Dichloromethane,
puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Expand
View Pricing
1.06054
Dichloromethane,
for gas chromatography ECD and FID SupraSolv
®
Expand
View Pricing
1.00668
Dichloromethane,
for gas chromatography MS SupraSolv
®
Expand
View Pricing
L090000
Dichloromethane,
Expand
View Pricing
24233
Dichloromethane,
puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Expand
View Pricing
1.06049
Dichloromethane,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, NF
Expand
View Pricing
8.22271
Dichloromethane,
EMPLURA
®
Expand
View Pricing
24233-M
Dichloromethane,
puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Expand
View Pricing
676853
Dichloromethane,
ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Expand
View Pricing
DX0835
Dichloromethane,
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
1.07020
Dichloromethane,
for analysis EMPARTA
®
ACS
Expand
View Pricing
Page 1 of 2
Page 1 of 2