Skip to Content
Merck
All Photos(1)

Documents

64306

Sigma-Aldrich

S-Methyl methanethiosulfonate

purum, ≥98.0% (GC)

Synonym(s):

S-Methyl thiomethanesulfonate, MMTS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2SCH3
CAS Number:
Molecular Weight:
126.20
Beilstein:
1446059
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

refractive index

n20/D 1.513 (lit.)
n20/D 1.513

bp

69-71 °C/0.4 mmHg (lit.)

solubility

chloroform: 750mg + 5 ml Chloroform mg/mL, colorless to light greenish-yellow

density

1.337 g/mL at 20 °C
1.337 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CSS(C)(=O)=O

InChI

1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3

InChI key

XYONNSVDNIRXKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Modification of Thiol Enzymes: S-methyl methanethiosulfonate (MMTS) offers a unique method for the modification of thiol enzymes and redox-regulated proteins, providing potential applications in biochemical research focused on enzyme regulation and redox biology (Makarov et al., 2019).

  • Sensor Development for Protease Activity: S-methyl methanethiosulfonate is used as a blocking reagent on the structural transitions of papain-like cysteine proteases, which supports its utility in sensor development, allowing for the detection and analysis of protease activity in various biological processes (Markovic et al., 2023).

  • Agricultural Pathogen Control: Research evaluating S-methyl methanethiosulfonate as a late blight inhibitor highlights its potential as a broad-range toxin against plant pathogens, suggesting applications in agriculture for the management of crop diseases (Joller et al., 2020).

Caution

may discolor to yellow on storage

Other Notes

Methylsulfenylating agent with many applications; For the α-methylsulfenylation of cyclic ketones; Methyl disulfides from thiols; selective, reversible inactivation of enzymes

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Laszlo et al.
The Journal of Organic Chemistry, 49, 2281-2281 (1984)
D. Scholz
Synthesis, 944-944 (1983)
W H Briggs et al.
Journal of agricultural and food chemistry, 48(11), 5731-5735 (2000-11-23)
Thiosulfinates (TSs) have been implicated as a principle source of the antiplatelet property of raw onion and garlic juice. The in vitro responses of human platelets to dosages of four TSs were measured using whole blood aggregometry and compared by
D J Smith et al.
Biochemistry, 14(4), 766-771 (1975-02-25)
New reagents for the temporary blocking of active or accessible sulfhydryl groups of enzymes have been developed. These reagents, which are either alkyl alkanethiolsulfonates or alkoxycarbonylalkyl disulfides, rapidly and quantitatively place various RS- groups on the sulfhydryls to generate mixed
Erin K Zess et al.
PLoS biology, 17(7), e3000373-e3000373 (2019-07-23)
Autophagy-related protein 8 (ATG8) is a highly conserved ubiquitin-like protein that modulates autophagy pathways by binding autophagic membranes and a number of proteins, including cargo receptors and core autophagy components. Throughout plant evolution, ATG8 has expanded from a single protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service