Skip to Content
Merck
All Photos(1)

Key Documents

63179

Sigma-Aldrich

3-(Maleimido)propionic acid N-hydroxysuccinimide ester

≥98.5% (HPLC)

Synonym(s):

3-(Maleimido)propionic acid N-succinimidyl ester, BMPS, N-(3-Maleimidopropionyloxy)succinimide, N-Maleoyl-β-alanine N′-hydroxysuccinimide ester, N-Succinimidyl 3-maleimidopropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2O6
CAS Number:
Molecular Weight:
266.21
Beilstein:
1492578
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.5% (HPLC)

form

solid

reaction suitability

reagent type: linker

mp

167-171 °C

functional group

NHS ester

storage temp.

2-8°C

SMILES string

O=C1CCC(N1OC(CCN2C(C=CC2=O)=O)=O)=O

InChI

1S/C11H10N2O6/c14-7-1-2-8(15)12(7)6-5-11(18)19-13-9(16)3-4-10(13)17/h1-2H,3-6H2

InChI key

JKHVDAUOODACDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Succinimidyl 3-maleimidopropionate (SMP, BMPS) is used as an aliphatic heterobifunctional crosslinker reagent. It is used for techniques such as the cross-linking of haptens to enzymes prior to enzyme immunoassays or fluorogenic substrates to peptidic antigens for use in fluorescence immunoassays.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Kitagawa
Enzyme Immunoassay, 81-81 (1981)
S J Xiao et al.
Journal of materials science. Materials in medicine, 8(12), 867-872 (2004-09-07)
Surface modification of acid-pretreated titanium with 3-aminopropyltriethoxylsilane (APTES) in dry toluene resulted in covalently bonded siloxane films with surface coverage that was relatively controllable by regulating the reaction conditions. A hetero-bifunctional cross-linker, N-succinimidyl-3-maleimidopropionate (SMP), reacted with the terminal amino groups
S Heyse et al.
Protein science : a publication of the Protein Society, 4(12), 2532-2544 (1995-12-01)
A new method is presented for measuring sensitively the interactions between ligands and their membrane-bound receptors in situ using integrated optics, thus avoiding the need for additional labels. Phospholipid bilayers were attached covalently to waveguides by a novel protocol, which
M C Durrieu et al.
Journal of materials science. Materials in medicine, 15(7), 779-786 (2004-09-28)
Ceramics possess osteoconductive properties but exhibit no intrinsic osteoinductive capacity. Consequently, they are unable to induce new bone formation in extra osseous sites. In order to develop bone substitutes with osteogenic properties, one promising approach consists of creating hybrid materials
Wuensch, E.
Hoppe-Seyler'S Zeitschrift fur Physiologische Chemie, 366, 53-53 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service