Skip to Content
Merck
All Photos(2)

Key Documents

P3415

Sigma-Aldrich

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester

≥95%, powder

Synonym(s):

N-Succinimidyl 3-(2-pyridyldithio)propionate, SPDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O4S2
CAS Number:
Molecular Weight:
312.36
Beilstein:
1548137
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NC.07

Assay

≥95%

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

84-86 °C (lit.)

solubility

acetone: 50 mg/mL
DMF: soluble

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(CCSSC1=NC=CC=C1)ON2C(CCC2=O)=O

InChI

1S/C12H12N2O4S2/c15-10-4-5-11(16)14(10)18-12(17)6-8-19-20-9-3-1-2-7-13-9/h1-3,7H,4-6,8H2

InChI key

JWDFQMWEFLOOED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester (SPDP) is a heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. It is typically coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thiol-disulfide exchange buffered at pH 8.0 (7.5-8.5). Alternately, this reagent may be used as a protected thiolating reagent following initial coupling and reduction of linker. It incorporates four atom linker.

Application

3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester (SPDP) is useful for the preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.
It has been used for antibody conjugation and as a protein cross-linker.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidenori Matsuo et al.
Journal of nanobiotechnology, 16(1), 59-59 (2018-08-06)
Various nanocarriers have been used to deliver subunit vaccines specifically to dendritic cells (DCs) for the improvement of immunogenicity. However, due to their insufficient DC priming ability, these vaccines could not elicit effective innate immunity. We have recently developed a
Integrin-independent tyrosine phosphorylation of p125fak in human platelets stimulated by collagen.
Achison M
The Journal of Biological Chemistry, 276, 3167-3174 (2001)
Protein Engineering for Therapeutics (2012)
Targeting chemotherapy using antibody-combined liposome against human pancreatic cancer cell-line.
Akaishi S
The Tohoku Journal of Experimental Medicine, 175, 29-29 (1995)
Bispecific Antibodies (2011)

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service