Skip to Content
Merck
All Photos(1)

Documents

62280

Supelco

Methyl linoleate

analytical standard

Synonym(s):

Linoleic acid methyl ester, Methyl cis,cis-9,12-octadecadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)2(CH2)7CO2CH3
CAS Number:
Molecular Weight:
294.47
Beilstein:
1727614
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.462 (lit.)
n20/D 1.462

bp

192 °C/4 mmHg (lit.)

mp

−35 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7-,11-10-

InChI key

WTTJVINHCBCLGX-NQLNTKRDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl linoleate is a methyl ester of the tri-unsaturated essential fatty acid, linoleic acid.
Methyl linoleate, non-conjugated, is more easily oxidized than conjugated methyl linoleate .

Application

The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation in vivo .
This analytical standard can also be used as follows:

  • Comparative analysis of gas chromatography-combustion-mass spectrometry and gas chromatography-flame ionization detector methods for the determination of fatty acid methyl esters (FAMEs) in biodiesel samples
  • Gas chromatography-tandem differential mobility spectrometry (DMS) based separation and quantification of 16 methyl- and ethyl- fatty acid esters from biodiesel samples
  • Simultaneous determination of fatty acid methyl esters in commercial food oil samples by gas chromatography-vacuum ultraviolet (GC-VUV) spectroscopy
  • Measurement of fatty acid methyl ester composition of various edible oil samples by 1H nuclear magnetic resonance (1H NMR) spectroscopy combined with partial least squares (PLS) method
  • Analysis of coffee oil and residue obtained from roasted coffee beans to determine the composition of 11 fatty acids following their methyl esterification by gas chromatography coupled with a flame ionization detector (GC-FID)
  • Simultaneous determination of fatty acids in bovine colostrum samples by GC-FID after their derivatization to ester forms using an acidic catalyst boron trifluoride

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Ying Feng et al.
European journal of medicinal chemistry, 46(4), 1198-1206 (2011-02-19)
In order to clarify the contribution of phenolic and enolic hydroxyl group to the antioxidant capacity of feruloylacetone, a model compound of half-curcumin, 6-(p-hydroxy-m-methoxyphenyl)-5-hexene-2,4-dione (FT), 6-(p-benzyloxy-m-methoxyphenyl)-5-hexene-2,4-dione (BMFT), 6-(m,p-dihydroxyphenyl)-5-hexene-2,4-dione (DDFT), 6-(p-hydroxy-m-methoxyphenyl)hexane-2,4-dione (DHFT), 6-(p-hydroxy-m-methoxyphenyl)-5-hexene-2,4-diol (THFT), and ethyl 2-(p-hydroxy-m-methoxybenzylidene)-3-oxobutanoate (EOFT) were synthesized. The
Guo-Xiang Li et al.
European journal of medicinal chemistry, 45(5), 1821-1827 (2010-02-04)
5,7-, 5,8-, 6,8-, 7,8-dichloro-4-quinolinol-3-carboxylic acid (5,7-, 5,8-, 6,8-, 7,8-DCQA) together with 7-chloro-4-quinolinol-3-carboxylic acid (7-CQA) and 4-quinolinol-3-carboxylic acid (QA) were synthesized to investigate the antioxidant properties. 5,7-DCQA exhibited the highest ability to scavenge 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cationic radical (ABTS+.), 2,2'-diphenyl-1-picrylhydrazyl (DPPH) and galvinoxyl
L Tesoriere et al.
Free radical research, 43(8), 706-717 (2009-06-24)
Reaction kinetics of betanin and its aglycone betanidin towards peroxyl radicals generated from the azo-initiated oxidation of methyl linoleate in methanol and of a heterogeneous aqueous/soybean phosphatidylcholine liposomal system were studied by monitoring formation of linoleic acid hydroperoxides and consumption
Roman Shchepin et al.
Journal of the American Chemical Society, 132(49), 17490-17500 (2010-11-26)
Free radical co-oxidation of polyunsaturated lipids with tyrosine or phenolic analogues of tyrosine gave rise to lipid peroxide-tyrosine (phenol) adducts in both aqueous micellar and organic solutions. The novel adducts were isolated and characterized by 1D and 2D NMR spectroscopy
Vitaly Roginsky
Chemistry and physics of lipids, 163(2), 127-130 (2009-11-03)
The kinetics of the chain free-radical oxidation of cardiolipin (CL), a unique phospholipid containing four linoleate moieties, have been studied for the first time. The technique based on monitoring oxygen consumption by using a Clark electrode was applied to determine

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service