Skip to Content
Merck
All Photos(1)

Documents

62210

Sigma-Aldrich

Methyl linolenate

technical, 70-80% (GC)

Synonym(s):

Linolenic acid methyl ester, Methyl cis,cis,cis-9,12,15-octadecatrienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7COOCH3
CAS Number:
Molecular Weight:
292.46
Beilstein:
1728194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

70-80% (GC)

form

liquid

refractive index

n20/D 1.470 (lit.)
n20/D 1.470

bp

182 °C/3 mmHg (lit.)

density

0.895 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h4-5,7-8,10-11H,3,6,9,12-18H2,1-2H3/b5-4-,8-7-,11-10-

InChI key

DVWSXZIHSUZZKJ-YSTUJMKBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iio et al.
Life sciences, 40(24), 2297-2302 (1987-06-15)
Secondary oxidative products of autoxidized methyl linoleate were divided into three groups (SP-I, SP-II and SP-III), which were then compared as to their abilities to form fluorescent substances and to degrade heme. SP-III showed a marked ability to produce two
William Y Boadi et al.
Journal of applied toxicology : JAT, 23(5), 363-369 (2003-09-17)
The single and combined effects of two abundant flavonoids, namely quercetin and genistein, were investigated according to their ability to inhibit the oxidation of methyl linolenate via Fenton's pathway. Antioxidative activity was determined by oxidizing methyl linolenate suspended in a
Patrícia Baptista et al.
Analytica chimica acta, 607(2), 153-159 (2008-01-15)
Biodiesel is the main alternative to fossil diesel. The key advantages of its use are the fact that it is a non-toxic renewable resource, which leads to lower emissions of polluting gases. European governments are targeting the incorporation of 20%
Rosario Zamora et al.
Journal of agricultural and food chemistry, 51(16), 4661-4667 (2004-01-07)
The oxidation of methyl linoleate (LMe) and methyl linolenate (LnMe) in the presence of bovine serum albumin (BSA) in the dark at 60 degrees C was studied to analyze the role of the type of fatty acid and the protein/lipid
Photoaddition of angelicin to linolenic acid methyl ester.
S Caffieri et al.
Journal of photochemistry and photobiology. B, Biology, 2(4), 515-521 (1988-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service