Skip to Content
Merck
All Photos(1)

Key Documents

60930

Sigma-Aldrich

Suberic acid

purum, ≥98.0% (T)

Synonym(s):

Octanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)6COOH
CAS Number:
Molecular Weight:
174.19
Beilstein:
1210161
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (T)

bp

230 °C/15 mmHg (lit.)

mp

140-144 °C (lit.)
140-144 °C

functional group

carboxylic acid

SMILES string

OC(=O)CCCCCCC(O)=O

InChI

1S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)

InChI key

TYFQFVWCELRYAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larisa Sheihet et al.
Biomacromolecules, 6(5), 2726-2731 (2005-09-13)
We describe the synthesis and characterization of a family of biocompatible ABA-triblock copolymers that comprised of hydrophilic A-blocks of poly(ethylene glycol) and hydrophobic B-blocks of oligomers of suberic acid and desaminotyrosyl-tyrosine esters. The triblock copolymers spontaneously self-assemble in aqueous solution
M Rivard et al.
Amino acids, 15(4), 389-392 (1999-01-19)
Papain-catalyzed regioselective cleavage of alpha-methyl ester in Z-DL-Asu(OMe)-OMe leads to Z-L-Asu(OMe)-OH and Z-D-Asu(OMe)-OMe. Subsequent saponifications yield Z-L-Asu-OH and Z-D-Asu-OH. The enzymatic alpha-ester hydrolysis was also achieved by subtilisin BPN' in organic solvent with low water content.
Monica Ilies et al.
Bioorganic & medicinal chemistry, 11(10), 2227-2239 (2003-04-26)
Novel matrix metalloproteinase (MMP)/bacterial collagenase inhibitors are reported, considering the sulfonylated amino acid hydroxamates as lead molecules. A series of compounds was prepared by reaction of arylsulfonyl isocyanates with N-(5H-dibenzo[a,d]cyclohepten-5-yl)- and N-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-yl) methyl glycocolate, respectively, followed by the conversion of
Q Gao et al.
Acta crystallographica. Section B, Structural science, 50 ( Pt 6), 695-703 (1994-12-01)
Neutron diffraction data for suberic acid [HOOC(CH2)6COOH] were collected at 18.4, 75 and 123 K using a twinned crystal. The neutron data reduction included derivation of a complete set of corrected intensities, as if from a single crystal. This was
Taishi Yokoi et al.
Dalton transactions (Cambridge, England : 2003), 41(9), 2732-2737 (2012-01-18)
Octacalcium phosphates (OCPs) co-incorporated with various molar ratios of succinate and suberate ions were synthesized by wet processing. The interplanar spacings of the (100) planes (d(100)) of OCPs formed in the presence of succinic acid (Suc) or suberic acid (Sub)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service