Skip to Content
Merck
All Photos(2)

Key Documents

P45001

Sigma-Aldrich

Pimelic acid

98%

Synonym(s):

Heptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)5CO2H
CAS Number:
Molecular Weight:
160.17
Beilstein:
1210024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

212 °C/10 mmHg (lit.)

mp

103-105 °C (lit.)

SMILES string

OC(=O)CCCCCC(O)=O

InChI

1S/C7H12O4/c8-6(9)4-2-1-3-5-7(10)11/h1-5H2,(H,8,9)(H,10,11)

InChI key

WLJVNTCWHIRURA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pimelic acid, also known as heptanedioic acid, is a dicarboxylic acid commonly used as a precursor in the biosynthesis of biotin. It serves as a building block in the synthesis of various compounds , and it is used as a key precursor in the production of polyamides, polyesters, and other polymers.

Application

  • Pimelic acid in microbial soil treatments: A 2023 study examined the use of pimelic acid in soil treatment, observing its impact on the microbial community structure and metabolites in bayberry plantations. This application illustrates its potential in agricultural biotechnology and soil remediation (Ren et al., 2023).
  • Pharmaceutical applications of pimelic acid: A 2023 study involved pimelic acid in the preparation and physiochemical analysis of novel ciprofloxacin/dicarboxylic acid salts, showcasing its utility as a pharmaceutical intermediate (Hibbard et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bio-based, amorphous polyamides with tunable thermal properties
Van Velthoven et al.
European Polymer Journal, 66, 57-66 (2015)
Kilogram Synthesis of Crebinostat
Yongjun et al.
Synthesis and Chemistry of Agrochemicals V, 48 (2016)
Pimelic acid, the first precursor of the B acillus subtilis biotin synthesis pathway, exists as the free acid and is assembled by fatty acid synthesis
Manandhar et al.
Molecular Microbiology, 104, 595-607 (2017)
B R Pettit
Clinica chimica acta; international journal of clinical chemistry, 156(1), 85-90 (1986-04-15)
A selected ion monitoring assay for thiodiglycollic acid in urine is described. Urine samples are analysed by combined gas chromatography-mass spectrometry as their dibutyl esters using pimelic acid as an internal standard. Rapid analysis was achieved by the simplification of
T Niwa et al.
Clinica chimica acta; international journal of clinical chemistry, 99(1), 71-83 (1979-11-15)
(1) 2,4-Dimethyladipic acid was first identified in normal human urine using gas chromatography-mass spectrometry. Urinary excretion of 2,4-dimethyladipic acid in 7 healthy adults ranged from 4.9 mumol to 14 mumol per 24 h. (2) Succinic acid, adipic acid, 3-methyladipic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service