Skip to Content
Merck
All Photos(1)

Key Documents

60357

Supelco

Potassium phthalate monobasic

reference material for titrimetry, certified by BAM, >99.5%

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

reference material

Quality Level

Assay

>99.5%

quality

certified by BAM

technique(s)

titration: suitable

pH

4

mp

295-300 °C (dec.) (lit.)

application(s)

environmental

format

neat

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydrogen phthalate, also known as KHP, is a volumetric standard produced under stringent conditions and measured with the highest possible precision. The content of the standard is measured using volumetric acid-base titrations. Its certified value and uncertainty measurement are in accordance with ISO Guide 35.

Application

It finds its use as a reference material for the standardization of volumetric solutions in alkalimetric titrations.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • High-purity material traceable to NIST SRM
  • High-quality offering accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Analysis Note

Exact concentration, expiry date and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purity of potassium hydrogen phthalate, determination with precision coulometric and volumetric titration--A comparison
Recknagel S, et al.
Analytica Chimica Acta, 599(2), 256-263 (2007)
Purity of potassium hydrogen phthalate, determination with precision coulometric and volumetric titration?A comparison
Recknagel S, et al.
Analytica Chimica Acta, 599, 256-263 (2007)
Acid- Base Dissociation Constants of o-Phthalic Acid in Acetonitrile/Water Mixtures over the (15 to 50)? C Temperature Range and Related Thermodynamic Quantities
Gagliardi GL, et al.
Journal of Chemical and Engineering Data, 55(1), 85-91 (2010)
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
H Fromme et al.
Environment international, 61, 64-72 (2013-10-10)
Phthalates have been used for decades in large quantities, leading to the ubiquitous exposure of the population. In an investigation of 63 German daycare centers, indoor air and dust samples were analyzed for the presence of 10 phthalate diesters. Moreover

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service