Skip to Content
Merck
All Photos(1)

Key Documents

42019

Supelco

Potassium dichromate

reference material for titrimetry, certified by BAM, >99.5%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

reference material

Quality Level

Assay

>99.5%

technique(s)

titration: suitable

mp

398 °C (lit.)

format

neat

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium dichromate is an oxidimetric standard produced under stringent conditions and measured with the highest possible precision. Its content determination and homogeneity assessment are performed using volumetric redox titration as the primary analysis method.

Application

It finds its use as a reference material for the standardization of volumetric solutions in redox titrations.

Features and Benefits

  • Available in a neat format in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Traceable to NIST SRMs
  • High-quality offering accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Analysis Note

Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in aquatic toxicity of potassium dichromate as a function of water quality parameters
Kikuchi M, et al.
Chemosphere, 170, 113-117 (2017)
Raman spectroscopic study of oxidation of aniline and N-methylaniline with dichromate
Mazeikiene R, et al.
Journal of Molecular Structure, 1139, 333-337 (2017)
Fu-Jun Liu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(2), 307-318 (2009-06-03)
Werner syndrome is a premature aging disorder characterized by cancer predisposition that is caused by loss of the Werner syndrome protein (WRN) helicase/exonuclease DNA repair protein. Hexavalent chromium is an environmental carcinogen and genotoxicant that is associated with respiratory cancers
Eduardo Molina-Jijón et al.
Toxicology, 291(1-3), 93-101 (2011-11-26)
Deferoxamine (DFO) is a recognized iron chelator which has been shown to exert nephroprotection in models of toxic nephropathies. In the present work the potential protective effects of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress were evaluated. Rats were injected
Bour-Jr Wang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 110(12), 775-779 (2012-01-18)
Occupational dermatitis among cement workers is a major occupational health concern. The two most important occupational hazards for cement workers are irritant and allergic cement contact dermatitis. The objective of this study was to investigate the severity of occupational cement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service