Skip to Content
Merck
All Photos(2)

Key Documents

49702

Supelco

Carnosol

analytical standard

Synonym(s):

Picrosalvin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26O4
CAS Number:
Molecular Weight:
330.42
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

Rosemarinus officinalis L. leaves

Quality Level

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

CC(C)c1cc2[C@@H]3C[C@H]4C(C)(C)CCC[C@]4(C(=O)O3)c2c(O)c1O

InChI

1S/C20H26O4/c1-10(2)11-8-12-13-9-14-19(3,4)6-5-7-20(14,18(23)24-13)15(12)17(22)16(11)21/h8,10,13-14,21-22H,5-7,9H2,1-4H3/t13-,14-,20+/m0/s1

InChI key

XUSYGBPHQBWGAD-PJSUUKDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carnosol is a naturally occurring phyto polyphenolor phenolic diterpene, found in rosemary, which can show a broad spectrum of applications as a an antioxidant, anticancer agent and anti-inflammatory agent.
Carnosol is one of the phenolic antioxidants present in extracts of the herb rosemary (Rosmarinus officinalis). Carnosol inhibits the formation of tumors derived using irritants such as TPA and DMBA in murine models of carcinogenesis.The effective dose reported is 1-10 μM topically or 6 μg/ml when added to supernatants of cultured cells.Addition of rosemary extracts to the diet as a 1% supplement by weight was also found to decrease the frequency of DMBA-DNA adducts.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A phenolic diterpene with antioxidant and anticarcinogenic activities.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: salvia

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carnosol: a promising anti-cancer and anti-inflammatory agent
Johnson.JJ
Cancer Letters, 305, 1-7 (2011)
Trude Rakel Balstad et al.
Molecular nutrition & food research, 55(2), 185-197 (2010-09-10)
Cytoprotective gene products, e.g. phase II - and antioxidant enzymes, are important in cellular redox homeostasis. A common feature of these genes is binding sites for transcription factor nuclear factor erythroid-2-related factor 2 (Nrf2), named electrophile response elements (EpREs) within
Marvin J Núñez et al.
Phytochemistry, 72(4-5), 385-390 (2011-02-15)
Two ent-rosane- (cuzcol, 1 and 6-dehydroxycuzcol, 2) and a abietatriene- (salvadoriol, 3) type diterpenoids have been isolated from Maytenus cuzcoina and Crossopetalum uragoga, respectively, along with five known diterpene compounds (4-8). Their stereostructures have been elucidated on the basis of
Jeremy J Johnson
Cancer letters, 305(1), 1-7 (2011-03-09)
The Mediterranean diet and more specifically certain meats, fruits, vegetables, and olive oil found in certain parts of the Mediterranean region have been associated with a decreased cardiovascular and diabetes risk. More recently, several population based studies have observed with
Sang Yong Kim et al.
Journal of agricultural and food chemistry, 58(3), 1543-1550 (2010-01-16)
Carnosol, a naturally occurring bioactive phenolic diterpene originating from rosemary and sage, has been shown to exert antioxidant and anti-inflammatory effects. This study examined possible protective effects of carnosol on sodium nitroprusside (SNP)-induced cytotoxicity in C6 glial cells. Carnosol (1-10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service