Skip to Content
Merck
All Photos(1)

Key Documents

00390580

Rosmarinic acid

primary reference standard

Synonym(s):

(R)-O-(3,4-Dihydroxycinnamoyl)-3-(3,4- dihydroxyphenyl)lactic acid, 3,4-Dihydroxycinnamic acid (R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O8
CAS Number:
Molecular Weight:
360.31
Beilstein:
2227587
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

171-175 °C (lit.)

storage temp.

−20°C

SMILES string

OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2

InChI

1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1

InChI key

DOUMFZQKYFQNTF-WUTVXBCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Biochem/physiol Actions

Rosmarinic acid has shown to contain antioxidant and anti-inflammatory activities. Possesses promising physiological actions related to cognitive performance, Alzheimer′s disease prevention, kidney disease treatment, cardioprotection and cancer chemoprevention.

Other Notes

This compound is commonly found in plants of the genus: melissa mentha salvia thymus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Airoldi et al.
Chemistry, an Asian journal, 8(3), 596-602 (2013-01-11)
Amyloid peptides, Aβ1-40 and Aβ1-42, represent major molecular targets to develop potential drugs and diagnostic tools for Alzheimer's Disease (AD). In fact, oligomeric and fibrillar aggregates generated by these peptides are amongst the principal components of amyloid plaques found post
Anja Klančnik et al.
PloS one, 7(12), e51800-e51800 (2013-01-04)
Campylobacter is a major foodborne pathogen and alternative antimicrobials are needed to prevent or decrease Campylobacter contamination in foods or food producing animals. The objectives of this study are to define the anti-Campylobacter activities of natural phenolic compounds of plant
Eun-Ju Yang et al.
Journal of cellular physiology, 228(5), 975-982 (2012-10-09)
High mobility group box 1 (HMGB1) protein is a crucial cytokine that mediates response to infection, injury, and inflammation. Rosmarinic acid (RA) is an important component of the leaves of Perilla frutescens and has neuroprotective, anti-microbial, anti-oxidant, and anti-cancer effects
Rodrigo Lucarini et al.
Pharmaceutical biology, 51(9), 1087-1090 (2013-06-07)
Despite several pharmacological applications of Rosmarinus officinalis L. (Lamiaceae), studies on its analgesic and anti-inflammatory properties have been scarce. The aim of this work was to use in vivo models to evaluate the analgesic and anti-inflammatory activities of the aqueous
Juan A Martín et al.
PloS one, 8(2), e56987-e56987 (2013-03-08)
Efforts to introduce pathogen resistance into landscape tree species by breeding may have unintended consequences for fungal diversity. To address this issue, we compared the frequency and diversity of endophytic fungi and defensive phenolic metabolites in elm (Ulmus spp.) trees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service