Skip to Content
Merck
All Photos(1)

Documents

49617

Supelco

Dihexyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-dihexyl ester, Di-n-hexyl phthalate, Phthalic acid dihexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45
Beilstein:
1886839
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.486-1.489

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O

InChI

1S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14H,3-8,11-12,15-16H2,1-2H3

InChI key

KCXZNSGUUQJJTR-UHFFFAOYSA-N

General description

Dihexyl phthalate, a phthalic acid ester, is generally found in food and beverage products. It is used in cosmetics, and as an additive and a plasticizer in plastic production and food packaging materials. Dihexyl phthalate is regarded as a common environmental contaminant.

Application

Dihexyl phthalate may be used as an analytical standard for the determination of the analyte in food samples, beverages, and bottled water by various chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

350.6 °F

Flash Point(C)

177 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved method for rapid detection of phthalates in bottled water by gas chromatography-mass spectrometry
Otero P, et al.
Journal of Chromatography. B, Biomedical Applications, 997(24), 229-235 (2015)
Simultaneous determination of 11 phthalate esters in bottled beverages by graphene oxide coated hollow fiber membrane extraction coupled with supercritical fluid chromatography
Lou C, et al.
Analytica Chimica Acta, 1007(24), 71-79 (2018)
Shao-Hui Zhang et al.
Environmental science and pollution research international, 25(16), 15934-15942 (2018-03-29)
Widespread phthalate exposure has been recently documented and is hypothesized to increase blood pressure (BP) in humans. However, current studies have provided inconclusive evidence for an association between phthalate exposure and BP. Human epidemiologic studies on the topic remain lacking.
Yang Hongjun et al.
Environmental monitoring and assessment, 185(10), 8489-8500 (2013-04-24)
The Yellow River Delta (YRD) is a typical agricultural and petrochemical industrial area of China. To assess the current status of phthalate esters (PAEs) of soil residues, soil samples (0∼20 cm) (n = 82) were collected in Bincheng District, at the geographic center
Yu Liu et al.
Journal of analytical methods in chemistry, 2013, 419349-419349 (2013-04-12)
The presence of phthalate esters (PAEs) in the environment has gained a considerable attention due to their potential impacts on public health. This study reports the first data on the occurrence of 15 PAEs in the water near the Mopanshan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service