Skip to Content
Merck
All Photos(1)

Documents

44263

Supelco

Diisopentyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-bis(3-methylbutyl) ester, Bis(3-methylbutyl) phthalate, Diisoamyl phthalate, NSC 17070, Phthalic acid diisopentyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O4
CAS Number:
Molecular Weight:
306.40
Beilstein:
2134957
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.485-1.489

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCCC(C)C)C1=CC=CC=C1C(OCCC(C)C)=O

InChI

1S/C18H26O4/c1-13(2)9-11-21-17(19)15-7-5-6-8-16(15)18(20)22-12-10-14(3)4/h5-8,13-14H,9-12H2,1-4H3

InChI key

JANBFCARANRIKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diisopentyl phthalate may be used as a reference standard in the quantification the analyte in polyvinyl chloride (PVC) medical devices by gas chromatography coupled with mass spectrometry (GC-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

330.8 °F

Flash Point(C)

166 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantification of 14 phthalates and 5 non-phthalate plasticizers in PVC medical devices by GC?MS.
Gimeno P, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 949-950, 99-108 (2014)
Gabriel Jiménez-Skrzypek et al.
Journal of chromatography. A, 1611, 460620-460620 (2019-10-28)
In this work, poly(dopamine)-coated magnetic nanoparticles (Fe3O4@pDA) have been used as sorbents for the magnetic dispersive solid-phase extraction (m-dSPE) of a group of 10 phthalic acid esters (dipropyl phthalate, DPP, dibutyl phthalate, DBP, dicyclohexyl phthalate, DCHP, bis(2-ethylhexyl)phthalate, DEHP, di-n-octyl phthalate
Álvaro Santana-Mayor et al.
Journal of chromatography. A, 1565, 36-47 (2018-06-25)
In this work, the suitability of Fe3O4 nanoparticles coated with reduced-graphene oxide as sorbent was evaluated for the extraction of a group of fourteen phthalic acid esters (i.e. benzylbutyl phthalate (BBP), bis-2-n-butoxyethyl phthalate (DBEP), dibutyl phthalate (DBP), diisobutyl phthalate (DIBP)
Álvaro Santana-Mayor et al.
Food chemistry, 312, 125798-125798 (2019-12-31)
In this work, a green, inexpensive, simple and fast deep eutectic solvent (DES)-based dispersive liquid-liquid microextraction was evaluated, for the first time, for the extraction of phthalates (i.e. benzylbutyl phthalate, diisobutyl phthalate, diisopentyl phthalate, di-n-pentyl phthalate, di-(2-ethylhexyl) phthalate, di-n-octyl phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service